PSB-ICSP-1205 AS-HV_Incorrect Product Key
Description:
A. Background
Thermo Fisher Scientific has initiated a containment action and recall for certain Thermo Scientific™ Dionex™ AS-HV Autosamplers (P/N 064051) and the main boards (P/N 064237) due to an incorrectly configured product key.
When the product key is set incorrectly, Chromeleon™ does not recognize the PCB board and therefore the instrument as an AS-HV module.
B. Symptom
When attempting to connect an affected AS-HV Autosampler to Chromeleon, the following error messages displayed: “Not a Dionex ASHV module.”
Environment:
AS-HV P/N064051
AS-HV main board P/N064237
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1205 AS-HV Incorrect Product Key.pdf | August 27, 2026 |