19 Vial AS AP rack not supported in Empower part number 074938
Issue
Customer wanted to configure this tray type, part number 074938, as a custom plate type in Empower software
Dimensions and coordinates are needed
Environment
IC using an AS-AP autosampler
Autosampler with 19 position vial, AS-AP 10 mL Vial Tray, P/N 074938
Empower
Resolution
This tray type is not supported due to the odd number of vials configuration and cannot be used in Empower
See images below.
As a solution, the 40 position tray should be sold with the AS-AP for use with Empower, P/N 074936, AS-AP 1.5/0.3 mL Vial Tray.
Attachment(s)
| File | Last Modified |
|---|---|
| PN 074936-40 position tray.JPG | March 10, 2023 |
| 19 vial tray.png | July 06, 2022 |
| Picture2 scehmatic 074938.png | July 06, 2022 |