Thermo Fisher Scientific
AS-DV Sampler Carousel Spinning and Vial Reading Errors
Issue
An AS-DV sampler experiences multiple issues, including continuous spinning of the carousel and failure to locate specified vial positions. These issues occur at various stages of the sequence (beginning, middle, or end). Error messages indicate that vials are not found in the carousel, and the system sometimes misidentifies vial sizes or fails to recognize vials. There have also been reports of the carousel spinning nonstop and unabel to locate the "home" position.
Environment
AS-DV autosampler
Resolution
1.      **Check and Tighten spindle assembly Set Screws**:
clipboard_e147cb2ed23f61e2cd10f3f6a2f8be8a0.png
-          Inspect the spindle assembly for loose set screws.
-          Tighten any loose set screws to ensure the spindle assembly is secure.
-          Ensure screws are aligned and tightened to the flat surfaces properly to prevent the spindle from shifting over time.
-          Refer to the AS-DV service manual for proper belt tension instructions.
2. **Level the Carousel:**
-          Ensure the carousel is spinning level.
-          Adjust the spindle assembly as needed to achieve proper alignment.
3. **Replace Spindle Assembly (if necessary):**
-          If aligning and adjusting the screws do not resolve the issue, replace the spindle assembly.
4. **Reassemble and Test:**
-          Reassemble the unit and align the belt of the carousel.
-          Run the unit to verify that the issue has been resolved.
5.  If these adjustments  don’t resolve the issues, consider replacing the following parts:
·       PCB ASSY, CPU, AS-DV-NPB (P/N 069204)
·       TRANSMITTER SENSOR BOARD (P/N 069317)
·       RECEIVER SENSOR BOARD (P/N 069315)
·       SPINDLE ASSEMBLY (P/N 071573)
Analysis
The issue may stem from a mechanical misalignment or loose components within the spindle assembly, causing the carousel to lose its correct orientation. Improper alignment and tightening of the pulley screws can lead to the spindle shifting over time, causing communication errors between the transmitter and receiver. There could also be a problem with the belt tension, so consider these possibilities before replacing parts.
Cause
The root cause of the issue can be from loose set screws and improper alignment within the spindle assembly, leading to misalignment and improper sensor operation. Ensuring that the spindle assembly and pulley are securely fastened and properly aligned can help maintain the correct operation of the carousel.
  • Was this article helpful?