AS-DV produces grinding noise or high pitched sound
Issue
AS-DV produces grinding noise or high pitched noise when loading vials. There may be a grinding or high pitch sound observed when the sample head is moving down due to lack of lubrication.
Environment
AS-DV Autosampler
Resolution
Lubricate guide rod with AS-DV grease (part number 076541):
Step 1: Lubricate the lead screw with grease (part number 076541).
Step 2: Lubricate the guide rod with grease (part number 076541).
Step 3: Run the sample head up and down a few times to distribute the lubricant evenly and this should eliminate the issue.
Step 4: Run 2-3 test injections of customer standards to verify the sampler works under normal operation conditions.
Review the attached video for information lubricating the head guide rod.
Cause
The lead screw normally gets lubricated but it has been discovered that a number of AS-DVs had a lack of lubrication on the sample head guide rod, which is positioned just behind the lead screw.
Attachment(s):
| File | Last Modified |
|---|---|
| AS-DV grinding.jpg | January 09, 2022 |