AS-DV produces high back pressure or damages caps
Issue
- AS-DV caps are damaged when sample is loaded.
- AS-DV produces grinding or high pitched noise while loading.
Environment
AS-DV Autosampler
Resolution
Replace sample tip and transfer line (part number 071575):
Step 1: If back pressure is suspected, first disconnect the transfer line from the AS-DV at the load/inject valve - this will isolate the AS-DV. Test the sampler and if no back pressure is observed, proceed to troubleshoot the load/inject valve and downstream IC system.
Step 2: If back pressure is observed in the AS-DV, replace the tip and transfer line - part number 071575 or perform a full PM on the AS-DV sampler. AS-DV Symptoms of back pressure isolated in the AS-DV include damaged vial caps and the cap filters being pushed out of the cap assembly.
Step 3: If back pressure is still observed or vial caps are still being damaged, troubleshoot the Load/Inject Valve or IC system.
Attachment(s)
| File | Last Modified |
|---|---|
| AS-DV high backpressure.jpg | January 09, 2022 |