AS AP coolant solution part number and solution
Issue
The part number for the AS AP coolant solution is not in the manual, it is in a separate PSB IC sys 1029
Environment
Ion Chromatography AS AP autosampler
Resolution
Part numbers: (photo needed)
082891 COOLANT,AS-AP,30%IPA,500ml
082907 KIT,ASSY,COOLANT REFILL,AS-AP,TEMP