Acquiring pH during runs using an electrochemical detector
Creating a virtual channel to collect pH trace during electrochemical detection
Issue
Collect pH trace during electrochemical runs
Environment
ICS
Electrochemical detection
Resolution
In order to collect the pH trace over a run, it is necessary to set up a Virtual Channel. Enter the command into the instrument method script to acquire pH over the course of an acquisition using electrochemical detection.