Change suppressor or suppressor not recognized error ADRS CDRS Chromeleon CMD suppressor type ERS configuration
Issue
Customer's current method will not work, they just changed their suppressor and receive an error about their DRS suppressor. Consumable Monitoring Disabled suppressors (CMD after the suppressor part number) cannot be used with their current instrument method and this must be modified otherwise ready check will fail or suppressor errors will be seen in Chromeleon.
Environment
ICS-6000
ICS-5000+
ICS-5000
Integrion
Inuvion
Aquion
Chromeleon
Resolution
The easiest solution is to uncheck then recheck the suppressor option in electrolytics, save the installation and then create a new instrument method.
The other option is when the ready check fails in Chromeleon to correct the two errors:
Click on the source of the error, Electrolytic Suppressor Type or Electrolytic Suppressor PowerMode and correct the errors in the script editor.
The suppressor type needs to be changed from ADRS to AERS in this case or from CDRS to CERS (select appropriate 2 or 4 mm).
The suppressor power mode line can be corrected or deleted. Save the Instrument Method changes and confirm the ready check passes so the new sequence can be run.
Cause
Attachment(s)
| File | Last Modified |
|---|---|
| clipboard_e98f2621c3d1469b96c8b78b81a46fa02.png | April 15, 2025 |
| clipboard_eb72087c6f790de70ac6256b5f9558b46.png | April 15, 2025 |
| clipboard_e2ab9378206c9a4e2ea372bb03d7a4e12.png | April 15, 2025 |
| clipboard_e07e7b4f4bc7f7568f8078af91913e001.png | April 15, 2025 |
| clipboard_ede289bec9a40959d59704ed7e8053517.png | April 15, 2025 |
| Customer Communication - CMD suppressors and columns without RFID tags.pdf | April 15, 2025 |