Changing AS-AP Loop
Issue
How do I change sample loop size in the configuration?
Environment
- AS-AP Autosampler
- Chromeleon 7
- Chromeleon 6.8
Resolution
| Navigate to instrument configuration | |
| Find the Chromeleon icon by clicking the arrow located at the bottom right of the comptuer screen. | ![]() |
| RIght-click on the Chromeleon icon and choose Configure instruments | ![]() |
| Expand all icons as seen under each instrument name. Double-click your sampler | ![]() |
| On the options schreen, change loop size 1 for system one and loop size 2 for system 2. For a single system, only loop size 1 is only available to edit. | ![]() |
| Click OK at the bottom and save the installation configuration on tool bar. | ![]() |
Attachment(s)
| File | Last Modified |
|---|---|
| sample_loop_figure03.png | January 09, 2022 |
| sample_loop_figure04.png | January 09, 2022 |
| sample_loop_figure05.png | January 09, 2022 |
| sample_loop_figure02.png | January 09, 2022 |
| sample_loop_figure01.png | January 09, 2022 |