Chromeleon 7.3.2 Thermo Scientific IC Drivers moved to “Obsolete” category
Issue
With the release of Chromeleon 7.3.2 the ICS5000+ / ICS PDA Photodiode Array Detector / ICS-900 / ICS-1100 / ICS-1600 / ICS-2100 and other additional instrument drivers have been moved into the Obsolete driver section of the software release. These specific instrument drivers, although not obsolete are discontinued. They can be found in the Installation Configuration Manager along with other legacy obsolete drivers.
Environment
Chromeleon 7.3.2 Thermo Scientific IC Drivers moved to “Obsolete” category.
Resolution
The"Obsolete" drivers have been moved into the Obsolete / Thermo Scientific IC category of the Chromeleon 7.3.2 Instrument configuration. The attached PDF has instructions on how to find them.
Attachment(s)
| File | Last Modified |
|---|---|
| Chromeleon 7.3.2 Thermo Scientific IC Drivers moved to “Obsolete” category.pdf | May 22, 2023 |