Thermo Fisher Scientific
Chromeleon Audit trail problems to fix system out of memory error
This articles includes information about how to troubleshoot and resolve a "system out of memory" error in the Chromeleon audit trail.
Issue
Audit Trail system out of memory error. 
Environment
Chromeleon 7.2.9
Resolution
Try steps:
1  run:  sfc/scannow   in cmd prompt as admin
2  execute station IQ and fix errors/ warning
3  ask level 2 support