DI Water Causing Extra Peaks, High Total Conductivity, Peaks Larger Than Expected
Issue
- IC system total conductivity is too high or noisy.
- Unknown peaks show up.
- Individual peaks are too large.
Environment
Ion Chromatography
Resolution
Step 1: Check DI Water System is functioning properly.
Step 2: Verify that the DI water system is meeting the following minimum criteria:
- Ions–Resistivity >18.0 (megohm-cm)
- Organics–TOC <10 ppb
- Iron/Transition Metals* <1 ppb
- Pyrogens <0.03 (Eu/mL)
- Particulates >0.2 μm <1 (units/mL)
- Colloids–Silica <10 ppb
- Bacteria <1 (cfu/mL)
* Iron/transition metal content not specified for ASTM Type I water
Step 3: If the DI Water system does not meet ALL criteria, the DI water being used is not suitable for Ion Chomatography and an alternate source of DI Water should be used.
Step 4: If the water system meets the DI Water specifications, then inject a DI Water blank to check for unknown or larger than expected known peaks in the DI Water.
Step 5: A peak or larger than expected peak in the DI Water indicates the water being used is not suitable for Ion Chomatography and an alternate source of DI Water should be used.
Attachment(s)
| File | Last Modified |
|---|---|
| DI Water System - image.png | January 11, 2022 |