Dionex Aquion Pump Is On but There Is Little or No Eluent Flow
Issue
The Dionex Aquion pump is turned on in Chromeleon, but normal eluent flow and/or system pressure is not produced.
The user may report one or more of the following:
- The pump shows ON, but little or no liquid appears to move through the system.
- The eluent valve does not appear to open when the pump is started.
- System pressure remains near zero or substantially below the normal operating pressure.
- The pump does not prime normally.
- Little or no liquid exits the pump prime/waste line when the priming valve is opened.
- The system previously operated normally and suddenly or progressively lost flow.
Note: Do not assume that the eluent valve has failed solely because the pump is ON and flow is absent.
Environment
- Thermo Scientific Dionex Aquion IC System
- Thermo Scientific Dionex Aquion RFIC System, where applicable
- Chromeleon CDS
Resolution
Before troubleshooting, record:
- Commanded pump flow rate
- Actual displayed pressure
- Expected normal pressure for the installed method
- Whether the problem is sudden, progressive, intermittent, or occurred after service
- Any recent tubing, pump, eluent, software, or maintenance changes
- Exact Chromeleon Audit Trail errors or warnings
- Whether liquid flows from the pump prime/waste line when the priming valve is opened
Recommended Troubleshooting Steps
-
Verify the pump command and flow setting in Chromeleon.
Confirm that the pump is commanded ON and that the requested flow rate is greater than zero.
For troubleshooting, use a known normal flow rate appropriate for the installed configuration, for example the customer's normal method flow.
Observe whether Chromeleon accepts the command and whether the displayed pump state follows the command.
-
Check the Chromeleon Audit Trail and instrument communication.
Review the Audit Trail for communication errors, device disconnects, pump errors, or other warnings.
If commands are not being accepted, the instrument repeatedly disconnects, or the displayed device state does not follow the commanded state, troubleshoot communication/configuration before assuming the eluent valve is defective.
-
Open the pump priming/waste valve and observe the prime waste line.
With the pump commanded ON, open the pump priming/waste valve and observe liquid flow to waste.
Interpret the result as follows:
- Strong, continuous liquid flow: Eluent is reaching the pump and the pump can move liquid. The eluent valve is unlikely to be the primary cause. Continue with the priming-valve and downstream-flow checks.
- Bubbles followed by normal flow: The pump or inlet line was likely air-bound or had lost prime. Continue priming until flow is bubble-free, then close the priming valve and retest.
- Little or no liquid flow: Continue troubleshooting the eluent supply, inlet tubing, eluent valve, pump inlet/check-valve path, or pump priming condition.
-
Verify the eluent supply to the instrument.
Confirm that:
- The eluent reservoir contains sufficient liquid.
- The pickup tube/filter is submerged.
- The inlet tubing is not kinked or pinched.
- No large air gap is present in the inlet tubing.
- The pickup/end-line filter is not visibly fouled or restricted.
If eluent does not freely reach the inlet side of the instrument, correct the reservoir, filter, tubing, or supply problem before evaluating the eluent valve.
-
Determine whether liquid reaches the inlet side of the eluent valve.
The purpose of this check is to separate an upstream supply problem from a valve/pump problem.
If liquid is not reaching the valve inlet, the eluent valve is not the root cause.
If liquid reaches the valve inlet normally but does not pass downstream when the pump is commanded ON and communication is normal, the eluent valve or its control becomes more likely.
-
Close the priming/waste valve and observe pressure response.
After obtaining bubble-free prime flow, close the priming valve finger-tight.
Observe system pressure.
- If pressure rises toward normal, the system had likely lost prime or contained air.
- If liquid continues flowing primarily to prime waste after the valve is believed to be closed, the priming/waste valve may not be sealing.
- If prime flow is strong but normal system flow or pressure is still absent after closing the valve, continue downstream.
-
Separate inlet/pump problems from downstream restrictions or leaks.
If the pump produces normal liquid flow to the prime waste line but does not produce normal flow or pressure through the analytical flow path, investigate:
- Priming/waste valve not fully sealing
- High-pressure leak
- Pump check-valve behavior
- Pump performance under load
- Downstream restriction
- Incorrect or recently changed plumbing
Use divide-and-conquer isolation rather than replacing components.
-
Measure actual flow when commanded flow is uncertain.
If practical and safe for the installed configuration, collect liquid over a known time at an appropriate outlet or diagnostic boundary and compare measured flow with the commanded flow.
A displayed flow setting alone does not prove that the hydraulic flow is correct.
-
Replace or service the eluent valve only after the fault has been isolated to that component.
The eluent valve becomes a leading suspect when:
- Eluent supply to the valve inlet is confirmed.
- Pump commands and communication are normal.
- The pump is commanded ON at a valid flow rate.
- The pump cannot establish normal inlet flow or prime because liquid does not pass the valve.
- Upstream reservoir/tubing restrictions and obvious air-lock conditions have been eliminated.
Follow the current Aquion-specific service documentation for valve replacement, electrical testing, or internal service procedures.
Analysis