Does the ICS-6000 pump motor truly stop when communication fails
Issue
Field reports indicate that the 'Keep pumping even if communication fails' option does not work properly when unchecked.
Environment
ICS-6000 DP
ICS-6000 SP
Resolution
User needs to wait for 5 minutes after the communication between ICS-6000 pump and Chromeleon fails when unchecking the "Keep pumping even if communication fails" in the pump module configuration settings
Analysis
Simulating the communication failure mode by completely removing the ICS-6000 pump USB cable from the rear panel
| Communication Failure mode | Checking the “Keep pumping even if communication fails” | Un-checking the “Keep pumping even if communication fails” |
| Unplugging the USB cable from the pump rear panel | Pump motor didn’t stop after 5 minutes | Pump motor stopped 5 minutes after unplugging the pump USB cable. |
Cause
By pump firmware. There is a timer in pump firmware to monitor the USB communication, whenever the timer times out, pump stops.
Exception: The pump will not stop if the pump firmware doesn't not detect the true communication failure.
- A test was done that by hitting the pump "Disconnet" button in the Chromeleon ePanel, it was observed the pressure can still change even the "connection" button is dark in Chromeleon.