EG/EGC Over Voltage Errors
Error Code
EG Over Voltage
EGC Over Voltage
(May be associated with excess gas formation/bubbles in the EGC)
Environment
IC models using eluent generation
ICS-6000/5000+
Integrion
Inuvion
Cause
While the cause can be a failure of the EGC, the most likely cause is the excessive formation of bubbles in the cell that are not dissipating properly. This can be caused by not enough backpressure on the system (among other causes, see also https://knowledge1.thermofisher.com/...rvoltage_error)
Resolution Steps
-
Shake the EGC to dislodge bubbles. Firmly tap the bottom of the EGC to continue to dislodge bubbles from the cell.
-
Confirm at least 2300psi of backpressure on the system (per the EGC operator manual).
-
If additional backpressure is needed, add restriction coil(s) between the eluent degasser and the injection valve to increase the system pressure.
-
If there is already >2300psi of backpressure on the system, add a black/red backpressure coil in line after EGC (black for standard bore flow rates (045877), red for micro bore flow rates (045878)).
-
Start the pump flow and allow the system to build pressure for a few minutes before turning on the EGC.
-
If it is not possible to operate the EGC after performing the steps above, replace the EGC.