Thermo Fisher Scientific
Enhancing the grounding of Inuvion system by a conductive waste tubing
Issue
For the Inuvion main board failure issue(KA article - Inuvion Error: The device could not be connected (The Leak Sensor is not present) ), ICSP R&D team confirmed that connecting a conductive waste tubing could help the instrument grounding which may avoid another main board failures.
Environment
Inuvion Core 
Inuvion 
Inuvion RFIC
Resolution
Install a conductive waste tubing to enhance static electricity discharge from the suppressor waste tubing. For example, use SST waste tubing or tubing similar to Integrion's conductive waste tubing (22153-60001).
22153-60001-2.png
For Inuvion CORE and Inuvion, connect the waste tubing 22153-60001 to the  Regen Out port of the Suppressor and direct the other end to the waste container bottom. 
For Inuvion RFIC, connect the waste tubing 22153-60001 to the Regen Out port of the CR-TC and direct the other end to the waste container bottom. 
 Connect Waste Tubing to CRTC.pngWaste Container-3.png
Connect the grounding wire of the waste tubing sheath to the lower right screw on the back panel. 
 Connect to chassis.JPG
  • Was this article helpful?