Example Standard Concentrations to Use for IC Anion, Cation Applications
Issue
I don't know what concentrations of standards to prepare when installing an anion or cation IC application using conductivity detection (CD).
Environment
Ion Chromatography
Resolution
Prepare standards that bracket the values on the column QAR (included with the analytical column). Examples:
|
Examples for Anions: |
||||||
|
P/N 057590 Stock (mg/L) |
1:10 dilution: 1ml stock diluted to 10ml |
1:20 dilution: 5ml of 1:10 dilution and dilute to 10ml |
1:100 dilution: 1ml of the 1:10 dilution and dilute to 10ml |
1:200 dilution: 5ml of 1:100 dilution and dilute to 10ml |
Example: AS18 QAR |
|
|
Fluoride |
20 |
2 |
1 |
0.2 |
0.1 |
0.5 |
|
Chloride |
100 |
10 |
5 |
1 |
0.5 |
3 |
|
Nitrite |
100 |
10 |
5 |
1 |
0.5 |
5 |
|
Bromide |
100 |
10 |
5 |
1 |
0.5 |
10 |
|
Nitrate |
100 |
10 |
5 |
1 |
0.5 |
10 |
|
Sulfate |
100 |
10 |
5 |
1 |
0.5 |
10 |
|
Phosphate |
200 |
20 |
10 |
2 |
1 |
N/A |
|
|
|
|
|
|
|
|
|
Examples for Cations: |
||||||
|
P/N 046070 stock (mg/L) |
1:10 dilution: 1ml stock diluted to 10ml |
1:20 dilution: 5ml of 1:10 dilution and dilute to 10ml |
1:100 dilution: 1ml of the 1:10 dilution and dilute to 10ml |
1:200 dilution: 5ml of 1:100 dilution and dilute to 10ml |
Example: CS12A QAR |
|
|
Lithium |
50 |
5 |
2.5 |
0.5 |
0.25 |
0.5 |
|
Sodium |
200 |
20 |
10 |
2 |
1 |
2 |
|
Ammonium |
250 |
25 |
12.5 |
2.5 |
1.25 |
2.5 |
|
Potassium |
500 |
50 |
25 |
5 |
2.5 |
5 |
|
Magnesium |
250 |
25 |
12.5 |
2.5 |
1.25 |
2.5 |
|
Calcium |
500 |
50 |
25 |
5 |
2.5 |
5 |