Full Uninstallation of Sii/Chromeleon 7
Issue
-
Fully uninstall Chromeleon 7
-
Fully uninstall SII
Environment
- Chromeleon 7
- SII
Resolution
Note: The following process will delete customer data. It is important to save or backup all data using the proper process prior to performing the following steps:
To completely remove Chromeleon 7 / SII (executables, program residue, and registry entries), first back up all important data using the Send To feature. Create a .cmbx file of the data for Chromeleon 7. It is not necessary to create a .cmbx file for SII.
-
Backup customer data and verify they have a copy that is accessible.
-
From C:\ProgramData\Dionex\Chromeleon directory, copy the InstrumentControllerConfiguration.cmic configuration file and the Chromeleon7_XXXX.cmlic configuration file. XXXX is the serial number.
-
Uninstall Chromeleon 7 by going to Control Panel, Program and Features, then Uninstall Chromeleon 7.
-
Delete the following directories:
-
C:\Program Files (x86)\Thermo\Chromeleon
-
C:\ProgramData\Dionex\Chromeleon
-
This will delete all data.
-
Remove the following registry entries:
-
HKEY_CURRENT_USER > Software > Dionex
-
HKEY_LOCAL_MACHINE > Software > Wow6432Node > Dionex
-
Remove the SQL by going to Control Panel, Program and Features, Uninstall SQL Server. This is usually SQL Server 2014 (64), but older versions may be 2008R2 or 2005.
-
Reboot computer.
The computer will then be ready for a fresh installation of Chromeleon 7 / SII. Remember all datavaults and data within will be deleted, so always safeguard and backup data before the procedure.