Thermo Fisher Scientific
How Do I Troubleshoot Liquid coming out of Suppressor ports
Issue: 
If there is liquid coming out of any of the four ports of the suppressor, what should I do?  
Environment: 
  • DRS 600
  • ERS 500e
  • AERS 500 Carbonate
Resolution: 
  1. Carefully tighten the fittings in the leaking port. 
  2. The fittings do not need to be more than 1/4-turn past finger tight.
  3. If tightening of the fittings does not stop the leak, replace the ferrules and bolts on the tubing. 
  • Was this article helpful?