How to understand the IC conductivity detector resolution system specification: 0.00238 nS/cm in Ion Chromatography instruments
Issue
Conductivity detector resolution:0.00238 nS/cm shows up in most ion chromatography system specification sheets and public documents. This detector resolution specification was written into commercial tenders and Thermo Scientific representatives were required to demonstrate or even test it during the installation.
Environment
Ion Chromatography instruments except ICS-600,ICS-900 and Easion.
Resolution
- Share the attached explanation letter to customers and explain that this 0.00238 nS/cm conductivity detector resolution is just a calculated specification which cannot be directly tested.
- Notify sales team and service team on how to understand and explain this specification.
- Advise sales team to add a note of "Non field tested specification" when the resolution has to be written into commercial tenders.
Cause
The conductivity detector design specification:Resolution 0.00238 nS/cm which is a calculated value was incorrectly interpreted as a field tested specification in tenders.
Attachment(s)
| File | Last Modified |
|---|---|
| Detector Resolution Explanation Letter.pdf | April 26, 2022 |