Thermo Fisher Scientific
ICS 5000 (plus) SP and DP pump head part numbers
Issue
How to identify the part number for the individual parts in and ICS-5000 or ICS-5000+ pump head. 
Environment
ICS-5000 SP/DP
Resolution
Primary Pump Head
ICS-5000 Primary Pump Head_1.jpg
1. Piston
P/N 068646 (Capillary)
P/N 079857 (Analytical)
2. Seal Retainer 
P/N 068627 (Capillary)
P/N 062092 (Analytical)
3. Piston Seal Wash Seal
P/N 068628 (Capillary)
P/N 063382 (Analytical)
4. Seal Wash Body
P/N 074446 (Capillary)
P/N 075767 (Analytical)
5. O-Ring
P/N 040695 (Capillary and Analytical)
6. Backup Ring
P/N 074371 (Capillary)
P/N 074370 (Analytical)
7. Main Piston Seal
P/N 075493 (Capillary)
P/N 075768 (Analytical)
8. Primary Pump Head
P/N 60-082602 (Capillary)
P/N 075047 (Analytical)
9. Outlet Check Valve Assembly 
P/N 044540 (Capillary)
P/N 045721 (Analytical)
10. Inlet Check Valve Assembly 
P/N 044541 (Capillary)
P/N 045722 (Analytical)
Secondary Pump Head
ICS-5000 Secondary Pump Head_1.jpg
1. Piston
P/N 068646 (Capillary)
P/N 079857 (Analytical)
2. Seal Retainer 
P/N 068627 (Capillary)
P/N 062092 (Analytical)
3. Piston Seal Wash Seal
P/N 068628 (Capillary)
P/N 063382 (Analytical)
4. Seal Wash Body
P/N 074446 (Capillary)
P/N 075767 (Analytical)
5. O-Ring
P/N 040695 (Capillary and Analytical)
6. Backup Ring
P/N 074371 (Capillary)
P/N 074370 (Analytical)
7. Main Piston Seal
P/N 075493 (Capillary)
P/N 075768 (Analytical)
8. Secondary  Pump Head
P/N 60-082661 (Capillary)
P/N 083482 (Analytical)
9. Priming Valve 
P/N 60-062087 
(Capillary and Analytical)
10. Priming Valve Seal
P/N 063382 
(Capillary and Analytical)