ICS modules won't connect to Chromeleon in Enterprise environment
Error Code
USB-XXXXXXXX not recognized
I/O operation aborted/ thread exits
Environment
ICS-6000
Integrion
This KA was created from multiple ICS6000 module cases in Enterprise environments but could potentially apply to other modules connected to Chromeleon.
Cause
This KA is also related to the Sentinel KA ICS-6000 DC Communications Disrupted by SentinelOne
The customer needs to be sure that the appropriate ports have been opened and exceptions have been allowed.
Resolution Steps
-
The recommended settings are found in the Chromeleon installation guide. This document is found on the computer with Chromeleon installed: Start Menu, All Apps/Programs, Thermo Chromeleon 7, Documents. It may be slightly different for different versions of Chromeleon.
-
Here's a snip of the TCP port settings (Firewall settings) from the installation guide for Chromeleon 7.3.2:
.