I am not seeing my panels, getting “$” instead of instrument name.
Issue
I am not seeing my panels, getting “$” instead of instrument name.
Environment
Chromeleon 7
Resolution
-
Select the epanel filter dropdown that says "last used"
-
Update the filter from "last used" to "autogenerated" by the drop down icon.
- Now the dollar signs should not be visible and you should have full control of your instrument.