Incorrect Peak Identifying Issue in Chromeleon - Chinese
Description:
This video illustrates that, when a component peak is incorrectly recognized or not recognized, Chromeleon’s output is N/A or other incorrect values. Comparing the retention times of the components in both standard samples and actual samples, and then adjusting the peak window by moving it forward or backward, could allow the peak name to be recognized and the component amount recalculated. In some situations, the peak window needs to be expanded to contain the desired peaks in both the standard samples and the actual analysis samples.
Environment:
- Chromeleon
- Ion Chromatography