Integrion : How to install and configure a manual injection valve
Issue
In some cases, customers use an autosampler and may need an additional manual injection valve to avoid cross contamination in the autosampler and the default injection valve.
This article describes how to connect a manual injection valve and how to configure the injection synchronization. with a minimum Instrument configuration modification.
Environment
Integrion
AS-AP or AS-DV autosampler
Manual Injection Valve
Chromeleon 7.2.10 or Higher
SII for Empower
Resolution
Injection valve installation :
Unscrew the Philips screw locking the bracket and move it at 90° :
Connect the manual injection valve on the bracket with the screw included in the bracket valve kit.
Use a flat washer (M8 – 25mm) between the screw and the bracket to properly tighten the valve.
Plumbing :
Connect the capillaries according to the following way :
o Connect sample loop between positions 1 and 4
o Connect pump to position 2
o Connect pre-column – column to position 3
o Connect waste to position 6
o Connect a no hole fitting to position 5. This position will be used to inject the sample.
Integrion TTL cable connection :
Connect the two black wires to the green connector included in the Integrion accessories kit.
o The first cable should be connected to TTL-1 OUT position
o The second cable should be connected to D-GND position
Chromeleon Instrument Configuration :
Open the Instrument Configuration Manager, select Inject Device Tab and uncheck InjectValve checkbox :
Do not include Remote Inject in Instrument configuration
Setting up the Instrument Method :
An Instrument method should be created specifically for the manual injection valve.
In the Scrip Editor, all the commands related to the AS-AP autosampler should be removed and 2 commands should be inserted in the start Run stage :
Wait Pump_ECD.TTL_Input_1.State=5v
WaitPump.ECD.TTL_Input_1.State=0v
Switching manually the injection valve will trigger the Acquisition
Analysis
Cause
Attachment(s)
| File | Last Modified |
|---|---|
| clipboard_e66d2b257049807085b2c61adfcb36d7f.png | March 11, 2025 |
| clipboard_e27a3b65aaba95fb7e394fca5e352d9c8.png | March 11, 2025 |
| clipboard_e26b9e65d4977b216b88a67853a5ae3db.png | March 11, 2025 |
| clipboard_e5812a3398e2dc2198763e1d6c2c011ec.png | March 11, 2025 |
| clipboard_eb37be60c810591ece473b0cd12b163f1.png | March 11, 2025 |
| clipboard_ed36cc40c4fb593b9966c9334471d7340.png | March 11, 2025 |
| clipboard_efabef9dc98b22c9ae9a2ae830d710e1f.png | March 11, 2025 |