Integrion high background conductivity and repeat CR-TC failure history resolved after suppressor replacement, with separate low-pressure alarm caused by eluent pickup above liquid level
Issue / Symptoms
The system showed repeated high background conductivity over multiple visits and repeated CR-TC failures over the prior six months. During follow-up cleaning with 100 mM KOH, conductivity continued rising above 20 µS, indicating the background issue remained active even after other flow-path work.
Environment
Product: PROD, INTEGRION, RFIC, DEGAS, OVEN, TEC
Serial Number: 20083072
Service Ticket: 4181127
Complaint Record: 5086234
Customer-Reported Presentation
The escalated follow-up consolidated repeated high background conductivity, multiple CR-TC failures over six months, occasional over-voltage conditions related to a very bubbly EGC, and later a low-pressure code during an overnight sequence. The complaint closure states the escalation was opened due to repeated high background conductivity and multiple CR-TC failures over the prior six months.
Cause
The best-supported root cause for the persistent high background was suppressor malfunction. The strongest evidence is that conductivity continued to rise above 20 µS during 100 mM KOH cleaning, the notes state this indicated the suppressor was not operating correctly, and conductivity settled to normal low values after replacing the ADRS 600 2 mm suppressor with an NGES-A-2 mm suppressor. A later Code 16 low-pressure event during the overnight run was separately traced to the eluent line sitting above the liquid level in the reservoir rather than to the suppressor itself.
Things Tried and Effectiveness
| Action Taken | Observed Outcome | Effectiveness Toward Primary Issue |
|---|---|---|
| Bypassed consumables and added a backpressure coil | Helped isolate the source of elevated conductivity and confirmed baseline performance under simplified conditions. | Diagnostic only |
| Changed EGC filters and removed air bubbles | Addressed over-voltage complaints and improved setup quality, but did not fully resolve the repeat high-background path. | Partial improvement |
| Replaced injection valve after rust-like buildup was observed behind the rotor seal | Corrected a separate contamination or wear finding, but high conductivity still persisted afterward. | Partial improvement |
| Replaced piston seals, rear seals, and O-rings on both pump heads | Performed during repeat troubleshooting, but conductivity still rose above 20 µS during 100 mM KOH cleaning. | No effect |
| Cleaned system with 100 mM KOH | Conductivity rising above 20 µS pointed toward suppressor malfunction. | Diagnostic only |
| Replaced ADRS 600 2 mm suppressor with NGES-A-2 mm suppressor | After hydration and equilibration, conductivity settled to about 0.770 µS and customer injections passed. | Confirmed corrective action |
| Corrected eluent reservoir pickup after Code 16 low-pressure fault | Pressure and conductivity returned to expected application values. | Confirmed corrective action for separate transient event |
Resolution
For persistent high background that remains elevated after flow-path checks and KOH cleaning, evaluate suppressor function directly. In this case, replacing the ADRS 600 2 mm suppressor with an NGES-A-2 mm suppressor resolved the high-background condition. If a later low-pressure alarm appears during the validation run, also verify that the eluent pickup remains below the liquid level in the reservoir before attributing that event to the original background issue.
Isolation / Verification
Only include steps reasonably supported by the evidence.
The team isolated conductivity behavior with consumables bypassed, verified proper detector-side pressure, corrected EGC bubbling issues, and then used 100 mM KOH cleaning to evaluate suppressor behavior. Continued conductivity rise above 20 µS supported suppressor malfunction. After suppressor replacement and hydration, conductivity settled to low values. Customer LCS injections passed, and after correcting the eluent pickup position, the system returned to expected operating pressure and conductivity.
Verification Outcome
The unit was documented as operating within specification, passing customer standards, and later behaving as expected at approximately 2318 psi and 0.631 µS after correction of the eluent reservoir issue. The complaint was closed as repaired and resolved, with continued monitoring noted because of repeat history.
Confidence
Issue definition: 5/5
Cause: 5/5
Resolution: 5/5
Isolation path: 4/5