Integrion unstable pressure and seal wash body leak with degas motor noise resolved after pump seal service and degas assembly replacement
Issue / Symptoms
System pressure cycled down from the normal range near 2100 psi to approximately 1700 psi and 300 psi, causing non-reproducible sample results. A small leak was present from the seal wash body vents, and abnormal cyclical squeaking was reported at startup.
Environment
Product: PROD, INTEGRION, RFIC, DEGAS, OVEN, TEC
Serial Number: 20083072
Service Ticket: 3146005
Purchase Order: 06182025LJC
Customer-Reported Presentation
The customer reported about one month of unstable pressure, inconsistent results, and a small leak from the seal wash body. They had already replaced multiple flow-path items, including tubing, suppressor, CRTC, seals, one piston, and some pump components, but the issue remained inconsistent. The customer also noted reddish residue near the secondary pump seal wash body vents and a startup noise that gradually went away after about 30 minutes.
Cause
The best-supported cause is a combination of leaking pump-head sealing components and a failed degas subsystem. The service report states the onsite leak was observed during operation, all pump seals and O-rings on both pump heads were replaced, and the degas vacuum pump and single-channel degas chamber were replaced after the vacuum pump was observed shutting itself off.
Things Tried and Effectiveness
| Action Taken | Observed Outcome | Effectiveness Toward Primary Issue |
|---|---|---|
| Customer replaced tubing, suppressor, CRTC, one piston, seals, check valves, and cleaned column and guard before field visit | Problem remained inconsistent before service visit. | No effect toward confirmed resolution |
| Field engineer observed system leak while operational | Leak condition confirmed onsite. | Diagnostic only |
| Replaced all pump seals and O-rings on both pump heads | System leak was reported as resolved after this repair. | Confirmed corrective action |
| Observed degas vacuum pump shutting itself off | Supported diagnosis of inadequate vacuum due to failed vacuum pump or breach in degas chamber. | Diagnostic only |
| Replaced degas vacuum pump and single-channel degas chamber | Degas function was reported fully restored successfully. | Confirmed corrective action |
| Customer performed overnight injections | Full functionality was reported restored. | Confirmed corrective action |
Resolution
Replace pump-head seals and O-rings to stop the operational leak, then replace the degas vacuum pump and degas chamber after the degas vacuum pump was observed shutting off and failing to establish vacuum properly. The report documents restored degas function and successful overnight injections afterward.
Isolation / Verification
Only include steps reasonably supported by the evidence.
Leak was confirmed while the system was running. The degas vacuum pump was also directly observed shutting itself off, which supported degas fault isolation. The customer then ran successful overnight injections after repairs.
Verification Outcome
Leak resolution and degas recovery were verified by the service engineer, and the customer completed successful overnight injections with functionality restored. The report marked the unit as operating as intended.
Confidence
Issue definition: 5/5
Cause: 4/5
Resolution: 5/5
Isolation path: 4/5