Inuvion Error: The device could not be connected (The Leak Sensor is not present).
Error Code
Inuvion: The device could not be connected (The Leak Sensor is not present).
Environment
Inuvion
Cause
There is a known issue with the early revision of the Inuvion main board that can cause FPGA chip burnout. This results in a "leak sensor is not present" error in the Chromeleon audit trail. If this is the case, the leak sensor is not the problem.
FPGA chip burnout issue symptoms:
- No LED lights are lit up after the instrument is powered up.
- Chromeleon can detect the Inuvion hardware, but when connecting the instrument, it will show “can’t connect with an error – the leak sensor is not present”.
- A red light on the board was lit up constantly(B).
- The large square FPGA chip(C) has burnt signs on the surface(not flat any longer). FSE can touch the chip surface with the finger while the instrument is off. (if there is no sign of burns on the chip surface, user can also feel the chip surface with the finger while the instrument is turned on, the chip surface is hot).
- Inuvion mainboard’s heartbeat LED(D) doesn’t flash(either off or constantly on).
- The red 10A fuse(A) on the filter may also be blown in some cases(but not all).
Damaged chip:
Factory has done some investigations about this issue, the suspect is that the +3.3V power input to the FPGA board can have large voltage surges during the instrument running, which exceeds the tolerance of the FPGA chip then caused the chip burnout. Based on the current reports and analysis, the mainboard was slightly re-designed on the layout of the mainboard and R&D added an LDO(Low-dropout regulator) to the 3.3V Input for the FPGA, improving FPGA internal PCI clamp diode by adding the TVS(Transient Voltage Suppressor) to protect the I/O pins against voltage overshoot.
The main board used in the current Inuvion production is Revision C, while the improved mainboard is Rev E. Testing of robustness continues of the Rev E version and stock is expected soon.
Resolution Steps
-
Confirm the symptoms are consistant with a damaged/burned FPGA chip.
-
Replace the main board of the Inuvion. (part 22185-61002R)