Thermo Fisher Scientific
Inuvion ship kit yellow tubing
Issue
The yellow (0.003" ID) and tan (0.007" ID) tubing included in the Inuvion ship kit l(22185-62000) look very similar in color. 
Environment
Inuvion
Resolution
Pay close attention to the ID of  tubing used when installing an Inuvion. In addition to red, black, green PEEK tubing, the ship kit, 22185-62000 also includes yellow and tan tubing.
The 0.003" ID (yellow) tubing is used to add additional backpressure between the RFIC degasser and injection valve (if needed, to achieve at least 2300psi).
The 0.007" ID (tan) tubing can be used if the Viper kit is not available, after the injection valve and can be used with either 2mm or 4mm column formats.  The column pre-heater (if installed) also has this 0.007" ID tubing installed.
clipboard_e7048e39e09cf8a92e98b6fec1e61f681.png
  • Was this article helpful?