Part number and replacement steps for the AXP pump check valve kit
Issue
What is the part number for the AXP pump check valve kit?
How do you replace the AXP pump check valves?
Environment
- AXP
- AXP-MS
Part Number
The part number for an AXP/AXP-MS pump check valve kit is C0960-01-00100 and the SAP description for this is: "KIT, PEEK, CHK VALVE,TELEDYNE, ROHS".
Resolution
The following steps are performed to replace the check valves on an AXP/AXP-MS pump:
1. Unscrew/uninstall the outlet check valve located at the top of the pump head.
2. Unscrew/uninstall the inlet check valve located at the bottom of the pump head.
3. Install new check valves and tighten firmly by hand Each check valve assembly contains two capsules. The sapphire seat in each capsule must be oriented downward in the final pump assembly.
Note: The inlet check valve has a larger opening (1/4”-28, flatbottom seat) for the 1/8-in. inlet tubing; the outlet check valve has a smaller opening (#10-32, cone seat) for the 1/16-in. outlet tubing. The inlet check valve must be connected at the larger opening in the pump head.
4. Reconnect all liquid lines and confirm that no leaks are present.
.png?revision=1)