Part numbers and assembly instructions for Integion manual injection tubing assembly
Issue
The part numbers 078493 and 60-078493 for the manual injection tubing assembly (SAP description: ASSY, TUBING,MAN INJ,ICS4) are no longer orderable. If the assembly is not available to order, the individual parts will need to be ordered and the assembly will need to be built.
What are the individual part numbers for the Integrion manual injection tubing assembly?
How to build the Integrion manual injection tubing assembly?
Environment
Integrion HPIC
Part Number
The part number for an Integion union is 039056 and the SAP description for this is: "FTG,UNION,1/4-28 ENDS,.06,P-1"
The part number for an Integion Luer adapter is 024305 and the SAP description for this is: "FTG,ADAPTR,1/4-28,FEM LUER"
The part number for an Integion 0.010IDX0.062OD PEEK tubing is 042690 and the SAP description for this is: "TUBING,0.010IDx0.062OD,PK,BLK,RoHS"
The part number for an Integion ferrule and lock ring is 062511 and the SAP description for this is: "FTG,FER,TFZ,+PK LK RNG,2PC,RoHS"
The part number for an Integion 1/4-28x1/16 nut fitting is 052230 and the SAP description for this is: "FTG,NUT,FLNGLS,1/4-28X1/16,PK,RoHS"
NOTE: When ordering part number 042690 order 12 inches for the quantity.
Resolution
To build the manual injection assembly, please follow these steps:
1. Assemble two piece ferrule and lock ring onto PEEK tubing flush as shown in photo below.
2. Screw the 1/4-28x1/16 nut fitting into coupler and tighten to set ferrule and ring onto tubing as shown in photo below.
Attachment(s)
| File | Last Modified |
|---|---|
| clipboard_e6aac0888a95a15523b731e0bf435068b.png | May 28, 2025 |