Thermo Fisher Scientific
Resolving Needle Arm Assembly Hitting 60mL Vials on ASE-350
Issue
We have found in some cases, not all, the sample compartment carousel is a little high, ~1 mm higher than normal. When running 60 mL sample vials, the needle arm assembly, particularly the sensor block, touches against the top of the vials as it moves in and out. This issue is not observed with the 250 mL bottles, as these bottles are a little shorter.  
Environment
ASE-350 
Resolution
I. Tighten Down the Cap
Since the gap between the vial cap and the sensor block is extremely small, ~1 mm, the sensor block can scape against the vial cap if the vial cap is loosely tightened.
II. Insert a Washer onto the Pivot Needle Block Rod
Another easy way to correct the issue is to raise the need arm assembly, ~1 mm, by inserting a washer onto the pivot needle block rod (#85).
III. Lower the Carousel
Remove the carousel and lower the bracket below it
Analysis
All the steps listed in the Resolution section were physically done. 
Cause
The sample compartment carousel is assembled a little high, ~1 mm higher than normal at the factory.
  • Was this article helpful?