Thermo Fisher Scientific
Sulfate Carryover Resolved by Replacing Tubing in Complete Flow Path
Sulfate Carryover Resolved by Replacing Tubing in Complete Flow Path
Issue
Customer reported persistent sulfate carryover and inconsistent sulfate QC performance on a Thermo Scientific Dionex Integrion HPIC system during low-level sulfate analysis. The issue persisted despite multiple troubleshooting attempts and component replacements.
Carryover was specifically observed during sulfate QC and blank injections, while nitrate injections remained stable during Basic Functionality Testing (BFT). The issue affected customer confidence in low-level sulfate quantitation and required extensive troubleshooting with Level 2 support.
Environment
  • Thermo Scientific Dionex Integrion HPIC System
  • AXP pump
  • External switching valve
  • Peristaltic pump
  • Concentrator method
  • External sample coil
  • Cation polisher
  • Low-level sulfate analysis
  • Customer site
Resolution
  1. Investigated persistent sulfate carryover following earlier troubleshooting adjustments and gathered chromatographic data for Level 2 support review.
  2. Performed isolation testing during blank injections by preventing the internal switching valve from actuating while external components were disconnected. No sulfate was detected during this condition.
  3. Performed standard blank injections with normal valve operation and observed sulfate carryover.
  4. Removed the peristaltic pump from the injection path while leaving the internal and AXP pumps active. Sulfate carryover remained present.
  5. Investigated the cation polisher as a possible contamination source.
  6. Removed the cation polisher from the system, but sulfate carryover persisted.
  7. Observed that allowing the internal valve to remain in the inject position for approximately one hour reduced sulfate carryover significantly.
  8. Modified method runtime from 12 minutes to 60 minutes to further evaluate cleanup behavior.
  9. Determined sulfate carryover was minimal with extended runtime, suggesting possible valve mechanism or pump-related issues.
  10. Replaced the injection valve mechanism and performed overnight testing.
  11. Observed stable injections without carryover following valve replacement, although response values remained elevated.
  12. Replaced the concentrator and sample transfer line.
  13. Performed customer QC injections and observed accurate data with no immediate carryover.
  14. Continued testing revealed sulfate carryover returned intermittently after initial success.
  15. Verified eluent water quality using a separate instrument and confirmed the water source was not contaminated with sulfate.
  16. Configured the system for a direct Basic Functionality Test per Level 2 guidance. Removed consumables, refreshed water supply, cleaned tubing, and installed a backpressure coil to maintain approximately 1600 psi.
  17. Allowed the system to flush overnight until background conductivity decreased below 1 µS.
  18. Performed BFT injections consisting of ten manual injections of 10 mg/mL nitrate standard at 1.0 mL/min and ten manual injections of 5 ppb sulfate standard per customer request.
  19. Submitted chromatographic data to Level 2 support for further analysis.
  20. Replaced all tubing directly in line with the sulfate QC flow path, including the external sample coil.
  21. Allowed the system to run overnight with repeated injections.
  22. Performed multiple QC injections and blank injections confirming stable sulfate response and absence of sulfate carryover.
  23. Customer completed additional sample analysis and post-sample QC testing confirming no sulfate carryover remained.
  24. Verified normal system operation was restored successfully.
Analysis
Troubleshooting focused on isolating sulfate contamination sources and determining whether the issue originated from the injection pathway, valve timing, pumps, consumables, or external hardware.
Testing demonstrated that nitrate performance remained stable during BFT testing while sulfate response was uniquely affected, suggesting sulfate-active contamination sites within the flow path. Extended inject-position dwell times reduced carryover, indicating adsorption and desorption behavior within the tubing or valve surfaces.
Removal of the cation polisher and verification of eluent water quality ruled out those components as contamination sources. Replacement of the valve mechanism improved consistency but did not permanently eliminate the issue. Final resolution occurred only after replacing all tubing in the sample flow path, indicating contamination or active sites throughout the sample flow path tubing assembly.
Cause
Sulfate-active contamination sites within the sample flow path tubing and associated transfer components caused intermittent sulfate carryover and inconsistent sulfate response. Complete replacement of all affected tubing eliminated the contamination source and restored normal system performance.
  • Was this article helpful?