The ED total background is high and there is high baseline noise. What can I do?
Environment
ICS-5000 ED Detector
ICS-6000 ED Detector
Resolution
Please ensure that the recommended 2M NaOH flush has been performed as described in the ED User Compendium. Failure to follow these recommendations may show symptoms like a high total background, and noisy baseline, due to potential biological growth in the system