Thermo Fisher Scientific
There is Leakage coming from the Side Seam of Suppressor
Issue: 
The suppressor is experiencing a leakage from the side seam. 
Environment: 
  • Dionex DRS 600
  • Dionex ERS 500e
  • Dionex ARES 500
Resolution: 
  1. Check for leakage from one of the four ports of the Dionex ERS 500 into the housing. 
  2. Check the back pressure after the suppressor.