There is no ion count left for the eluent cartridge
With a current or new EGC there could be an error in the chromeleon audit trail saying there is no communication with the EGC and also :There is no ion count left for the eluent cartridge. This cartridge can no longer be used. Please order replacement cartridge
Issue
Error message : There is no ion count left for the eluent cartridge. This cartridge can no longer be used. Please order replacement cartridge
Environment
IC system using EGC
Resolution
Remove the EGC serial number from the registry in Chromeleon 7, follow these steps:
-
Open the Registry Editor:
- On Windows 7, click Start and search for "regedit". Open regedit.exe .
-
Navigate to the Registry Location:
- For Chromeleon 7 in Windows XP:
- Go to
HKEY_LOCAL_MACHINE\SOFTWARE\Dionex\Chromeleon\Drivers\EG40\Cartridges.
- Go to
- For Chromeleon 7 in Windows 7:
- Go to
HKEY_LOCAL_MACHINE\SOFTWARE\wow6432Node\Dionex\Chromeleon\Drivers\EG40\Cartridges
- Go to
- For Chromeleon 7 in Windows XP:
-
Delete the Serial Number:
- There will be a separate folder for each eluent generator cartridge (EGC) under the Cartridges folder.
- Delete all of the folders corresponding to the old EG cartridge serial numbers