Troubleshoot AS-AP Run Completes But No Analyte Peak Is Observed After Injection
Issue
A Dionex AS-AP completes an injection or sequence without an obvious abort, but the chromatogram contains no expected analyte peaks.
The baseline and chromatographic run may otherwise appear normal, creating suspicion that the sample was not successfully drawn, transferred to the sample loop, or injected into the IC flow path.
Environment
- Thermo Scientific Dionex AS-AP Autosampler
- Chromeleon CDS
Resolution
Before troubleshooting, identify:
- AS-AP configuration: Push, Pull, Push Sequential, or Push Simultaneous
- Injection mode: full-loop, partial-loop, or other applicable AS-AP mode
- Installed sample-loop volume
- Syringe and buffer-line configuration
- Chromeleon version when command or configuration behavior is in question
- Whether tubing, loop, needle, syringe, valve, or sampler configuration was recently changed
NOTE: The AS-AP uses a syringe, syringe valve, sampling needle, buffer line, sample-transfer path, and injection valve to acquire and introduce sample. The exact liquid path depends on whether the sampler is configured for Push or Pull operation
Recommended Troubleshooting Steps
-
Use a fresh, easily detected standard.
Run a standard that normally produces clear analyte peaks. Avoid beginning the test with a complex customer sample so sample matrix or preparation does not complicate the diagnosis. -
Review the Chromeleon Audit Trail around the injection.
Look for warnings, errors, or unexpected commands near the injection time. Do not assume that a completed chromatographic run proves that sample introduction occurred successfully.Pay particular attention to messages involving:
- vial or tray position
- sample height
- injection-port alignment
- syringe condition
- syringe valve operation
- loop/injection-volume mismatch
- high-pressure valve switching
-
Confirm the configured AS-AP injection architecture.
Determine whether the method uses:- Push or Pull
- full-loop or partial-loop operation
- a single valve or additional AS-AP valve configuration
Do not interpret the sample path until the actual injection mode is known.
-
Prime and inspect the syringe.
From direct control/ePanel, prime the syringe and observe its operation.Verify:
- the plunger moves normally
- the syringe fills and empties
- no persistent air bubbles are present
- no liquid leaks from the syringe or syringe valve
- fluid movement is visible through the associated tubing
Continue priming until air is removed before evaluating injection performance. Persistent air, failure to fill, or leakage moves suspicion toward the syringe, syringe valve, wash supply, or associated tubing. 065437_AS-AP_Service Manual_Rev…
-
Verify the needle reaches the correct sample.
Observe one injection or controlled needle movement.Confirm:
- the carousel moves to the intended vial/well
- the configured tray matches the tray physically installed
- the needle moves to the expected location
- the needle enters the sample correctly
- sufficient sample volume is present
- sample height is appropriate
- the needle is not bent, damaged, or visibly misaligned
-
Inspect the buffer line and associated low-pressure sample path.
Check for:- trapped air
- empty tubing where liquid should be present
- loose fittings
- leakage
- kinks or restrictions
- incorrect tubing or syringe/buffer-line combination
Run a manual wash if necessary to confirm that wash liquid can move through the expected AS-AP path.
-
Verify the physical sample-loop size against Chromeleon configuration.
Confirm the actual installed loop volume and compare it to the loop size configured in Chromeleon.For full-loop operation, also verify that the injection volume requested by the method is appropriate for the installed loop.
A physical loop/configuration mismatch can prevent the intended injection sequence from operating correctly.
-
Inspect the sample-transfer line.
For Push configurations especially, inspect the tubing between the AS-AP injection port and the injection valve.Check for:
- leakage
- air
- disconnection
- incorrect routing
- kinks or restriction
- recent tubing replacement
If the Push-mode sample-transfer line was replaced or its length changed, verify that the applicable sample-transfer-line volume configuration/calibration has been handled according to the AS-AP procedure. Man-IC-AS-AP-Ops-July2025-DOC06…
-
Command the injection valve manually.
Use direct control to command the applicable valve between Load and Inject positions.Confirm that:
- Chromeleon reports the expected valve position
- the valve physically switches
- no switching error or timeout is recorded
The sample can be successfully drawn and transferred yet still fail to reach the column if the loop is never switched into the analytical flow path.
-
Compare direct/manual control with the automated sequence.
Perform the simplest supported injection operation using direct AS-AP/ePanel control where practical, then compare it with the customer's sequence.Interpret the result as follows:
- Direct/manual operation succeeds but sequence fails: prioritize Chromeleon method, sequence, configuration, or command timing.
- Direct/manual operation also fails: prioritize the physical sample-delivery or injection path.
- Syringe does not move liquid: prioritize syringe, syringe valve, air, wash supply, or tubing.
- Sample delivery appears normal but valve does not switch: prioritize valve control/configuration or valve hardware.
- Valve switches normally but no sample reaches the chromatograph: continue isolating the loop and transfer path.
-
Review the actual injection commands in the Instrument Method.
Do not rely only on the graphical method setup.Verify that the commands appropriate to the configured Push or Pull mode occur in the required sequence and that the injection valve reaches the appropriate Load and Inject states.
A run can continue chromatographically even if the intended sample-introduction operation is incomplete or misconfigured.
-
Perform a manual fixed-loop isolation test if the cause remains unclear.
Where the IC configuration permits, bypass the AS-AP sample-delivery path and manually overfill an appropriate fixed sample loop.Run replicate manual injections of the same standard.
- Manual fixed-loop injections produce normal peaks: move suspicion upstream toward the AS-AP syringe/needle/transfer path, sampler configuration, or commands.
- Manual fixed-loop injections also produce no analyte peak: keep the IC injection valve, column/separation, suppression, detector, and method branches open.
Do not replace AS-AP components until this fault boundary has been established when the manual isolation test is practical.
-
After correction, verify the original method.
Return the system to the customer's normal AS-AP method and confirm:- expected analyte peaks are present
- peak area is reproducible
- retention time is normal
- no new Audit Trail errors are generated
- the AS-AP completes repeated injections successfully
Analysis