Troubleshoot Degasser Malfunction Error Code 35 on a Dionex ICS-5000 Plus Single or Dual Pumps
Issue
The Dionex ICS-5000+ SP or DP reports:
Code 35 – “Degasser malfunction”
The error may occur during startup or operation and can remain active even though the vacuum pump can be heard running.
NOTE: Do not assume that Code 35 means the vacuum pump has failed. The degasser system includes the vacuum pump, vacuum tubing and connections, degas chamber, associated cabling/control, and vacuum-sensing function. A leak anywhere in the vacuum circuit can prevent the system from reaching the required vacuum condition.
Environment
- Product: Thermo Scientific Dionex ICS-5000+ SP
- Product: Thermo Scientific Dionex ICS-5000+ DP
- Software: Thermo Scientific Chromeleon CDS
Resolution
NOTE: The installed degasser configuration should be confirmed before ordering replacement components. ICS-5000+ configurations can use different degas chambers depending on the pump configuration.
Recommended Troubleshooting Steps
-
Record the exact failure conditions before replacing parts.
Record:
- ICS-5000+ SP or DP
- Pump configuration
- Isocratic or gradient configuration
- Exact Chromeleon Audit Trail message
- Whether Code 35 occurs immediately at startup or after operating for a period of time
- Whether the failure is constant or intermittent
- Whether the vacuum pump can be heard running
- Any recent maintenance, tubing changes, or degasser service
-
Command or observe operation of the degasser and determine whether the vacuum pump runs.
If the vacuum pump does not run, investigate the electrical/control branch first, including:
- Vacuum pump connection
- Degasser wiring/cabling
- Vacuum pump
- Associated control electronics
Do not replace the degas chamber solely because Code 35 is present.
-
If the vacuum pump runs, inspect the complete vacuum tubing path.
Inspect the tubing from the degas chamber to the vacuum pump for:
- Disconnected tubing
- Loose connections
- Cracks or splits
- Kinks
- Tubing deformation
- Poorly seated fittings
- Evidence of liquid or condensation in the vacuum line
Correct any obvious tubing or connection problem and retest the degasser.
-
Determine whether the vacuum pump can generate vacuum independently of the degas chamber.
Use the applicable service procedure and safe service access to isolate the vacuum pump from the chamber.
Interpret the result as follows:
- Pump cannot generate vacuum when isolated: the vacuum pump or its immediate connection/control becomes the primary suspect.
- Pump generates vacuum when isolated, but vacuum is lost when the chamber is reconnected: move suspicion toward the degas chamber, vacuum tubing, fittings, or chamber connections.
This is the highest-value isolation test because it separates the vacuum-generating component from the vacuum load.
-
If the pump passes the isolation test, inspect the degas chamber and its connections.
Check for:
- Damaged chamber connections
- Loose vacuum connections
- Physical damage
- Evidence of leakage
- Incorrect tubing installation
- Problems introduced during recent maintenance
Verify that the chamber installed matches the actual ICS-5000+ pump configuration before replacing it.
-
Evaluate intermittent behavior separately.
If Code 35 occurs intermittently, gently inspect the vacuum tubing and electrical connections for a condition that changes with movement or vibration.
Intermittent faults increase suspicion of:
- Marginal tubing connections
- Small vacuum leaks
- Loose electrical connectors
- Intermittent pump operation
- Vacuum-sensing/control problems
-
Evaluate the vacuum-sensing/control branch only after the vacuum system has been proven.
If:
- the vacuum pump runs,
- the pump can generate vacuum,
- the tubing and chamber hold vacuum,
- and Code 35 remains,
then move suspicion toward the vacuum-sensing or control system.
Do not condemn the sensor or control electronics until the physical vacuum circuit has been verified.
-
Verify the repair under normal operating conditions.
After correcting the fault:
- Reconnect the normal degasser plumbing.
- Start the instrument normally.
- Confirm that Code 35 does not return.
- Confirm that the vacuum pump behaves normally.
- Allow the system to operate long enough to verify that the degasser remains stable.
- Review the Chromeleon Audit Trail for recurring degasser errors.
Analysis