Troubleshoot Persistent High Background or Contamination on a Dionex ICS-5000+ Conductivity System
Issue
A Dionex ICS-5000+ conductivity system continues to show elevated background conductivity, contamination, ghosting, or an abnormal baseline after the system has already been flushed or cleaned.
The source of contamination is unclear and may be associated with one or more of the following:
- DI water
- manually prepared eluent or eluent reservoir
- reservoir pickup tubing or inlet hardware
- eluent generator or associated RFIC components
- mixer or other eluent-conditioning component
- autosampler/sample-introduction path
- guard or analytical column
- suppressor or regenerant path
- conductivity detector cell
- tubing or fittings elsewhere in the flow path
Environment
- Product: Thermo Scientific ICS-5000+ IC System
- Product: Thermo Scientific ICS-5000+ DC System
- Software: Thermo Scientific Chromeleon CDS
Resolution
Recommended Troubleshooting Steps
-
Confirm whether the problem occurs without an injection.
Run the system under normal analytical flow with no sample injection and allow it to equilibrate.Record:
- background conductivity
- pressure
- suppressor state
- temperature
- whether the method is isocratic or gradient
For an isocratic method, a reasonably stable baseline is expected after equilibration. For a gradient method, baseline movement may be normal; evaluate whether the gradient baseline profile is reproducible rather than expecting a flat baseline. Integrated_IC_Expert_Master_Dir…
-
Test with fresh, known-good Type I DI water.
Use fresh ASTM Type I water in a clean reservoir and avoid simply topping off an existing bottle.Inspect or clean as appropriate:
- eluent reservoir
- bottle cap
- pickup tubing
- pickup filter
- inlet fittings
Thermo documentation specifies Type I water for IC use and emphasizes low ionic and organic contamination. Man-IC-AS-AP-Ops-July2025-DOC06…
-
Determine whether the autosampler is involved.
If the high background is present continuously before any sample is injected, the autosampler is lower probability.If the baseline is clean before injection but contamination appears after a sample or standard is introduced, evaluate:
- sample carryover
- wash liquid
- needle/sample-transfer path
- injection loop
- sample matrix
Run a sequence such as:
water blank → high standard → water blank
A blank that becomes contaminated only after the high standard supports a sample-introduction or carryover source.
-
Simplify the conductivity flow path.
Using the applicable supported plumbing configuration, bypass as many upstream analytical components as practical and test the conductivity detector with clean water.A useful isolation concept is to remove or bypass, where applicable:
- EGC
- CR-TC
- mixer
- column
- suppressor
- CRD
The current Integrated IC OQ procedure uses this type of simplified detector-path configuration for conductivity testing by bypassing major analytical components and pumping water through the detector after equilibration. [Procedures]CDS OQ Chromeleon A…
-
Interpret the simplified detector-path result.
If the background remains high with clean water and the simplified detector path:
- suspect conductivity cell contamination
- immediate cell inlet/outlet tubing
- bubbles in the cell
- downstream restriction
- detector-side plumbing
- detector electronics only after fluidic causes are excluded
If the detector-only background is normal:
- the contamination is upstream
- restore components one at a time
-
Restore the suppressor and retest.
Reintroduce the suppressor while leaving other suspect components bypassed if the configuration allows.If the background becomes abnormal only after the suppressor is restored, investigate:
- suppressor operating state
- correct suppressor chemistry
- suppressor current/voltage as applicable
- regenerant or external-water flow
- bubbles or outgassing
- downstream backpressure or restriction
- suppressor condition
Do not replace the suppressor yet if a downstream restriction has not been excluded. The Integrated IC troubleshooting rules specifically treat suppression efficiency, regeneration, bubbles, detector-cell condition, and downstream backpressure as related variables. Integrated_IC_Expert_Master_Dir…
-
Restore the guard and analytical column and retest.
If the background becomes abnormal only after the column set is returned to the flow path, evaluate:- guard contamination
- analytical column contamination
- retained matrix material
- inappropriate cleaning history
- column-generated ghost peaks
Also inspect whether the symptom is:
- a constant conductivity offset
- discrete ghost peaks
- retention/selectivity change
- peak broadening
-
Restore RFIC or eluent-conditioning components individually.
If the system contains an EGC, CR-TC, mixer, CRD, EPM, or other eluent-generation/conditioning component, restore one component at a time.After each change:
- allow adequate equilibration
- record conductivity
- record pressure
- compare the baseline with the previous configuration
The first component whose addition causes the abnormal background to return becomes the primary branch for further troubleshooting.
-
Differentiate water contamination from reservoir/eluent-path contamination.
Compare:- fresh DI water directly from a known-good source
- fresh DI water in a clean reservoir
- the customer's normal reservoir and pickup hardware
- freshly prepared analytical eluent
If the problem follows the reservoir but not the source water, suspect contamination introduced after water collection.
-
After identifying the contaminated branch, follow the exact component-specific Thermo cleaning or service procedure.
Do not improvise cleaning chemistry or backflush direction for:- conductivity cells
- suppressors
- EGC/CR-TC components
- columns
-
Verify the repair under the original analytical method.
After the source is corrected, restore the complete system and confirm:- normal background conductivity
- stable baseline
- normal pressure
- normal retention
- acceptable response
- no carryover or ghosting
- acceptable repeatability
Analysis