Troubleshoot a Dionex ICS-6000 Analysis That Runs but Produces No Chromatographic Peaks
Issue
A Thermo Scientific Dionex ICS-6000 starts or completes an analysis, but no expected chromatographic peaks are observed.
The chromatogram may show a normal-looking baseline with no analyte peaks, or the detector signal itself may be abnormal or absent.
Environment
- Product: Thermo Scientific Dionex ICS-6000 Ion Chromatography system
- Software: Thermo Scientific Dionex Chromeleon CDS
Resolution
Before troubleshooting, identify:
- Exact ICS-6000 module and channel configuration
- Detector type
- Autosampler type
- Injection valve used by the affected channel
- Isocratic versus gradient method
- Column and guard column
- Eluent chemistry and concentration or eluent-generator program
- Suppressor type and operating mode, if applicable
- Method flow rate
- Expected and actual system pressure
- Detector background or conductivity
- Exact Chromeleon Audit Trail messages
- Chromeleon version
- Whether the problem began suddenly, progressively, after maintenance, after replumbing, or after a method/software change
Recommended Troubleshooting Steps
-
Review the raw detector signal before changing hardware.
Open the chromatogram and determine whether:
- a normal detector baseline is present but analyte peaks are absent, or
- the detector signal itself is abnormal, flat, missing, excessively high, or otherwise unusable.
If a normal baseline is present, initially prioritize sample delivery, injection, valve operation, and method commands.
If the detector signal is abnormal, include detector, suppression, bubbles, cell condition, and analytical flow in the initial fault tree.
-
Review the Chromeleon Audit Trail and Instrument Method.
Confirm that the expected modules are connected and ready.
Review the Instrument Method and verify that it contains the required commands for the affected channel, including:
- pump flow
- detector acquisition
- correct detector channel
- injection-valve position
- autosampler/sample-loading commands
- suppressor or electrolytic-device commands, where applicable
- eluent-generator commands, where applicable
- appropriate run time
Verify that the sequence is using the intended Instrument Method, vial position, injection settings, and instrument/channel.
-
Verify pump flow and pressure during the run.
Compare actual pressure with the historical pressure normally observed for the method.
If pressure is absent, abnormally low, unstable, or inconsistent with normal operation:
- verify eluent supply
- verify pump prime
- inspect for leaks or air
- confirm commanded flow
- measure collected flow when necessary
Resolve a significant flow-delivery problem before troubleshooting sample injection.
-
Confirm that liquid is flowing through the intended analytical path.
Verify the tubing path from the pump through the appropriate injection valve, column, suppressor where installed, detector, and waste.
Check especially for:
- recent plumbing changes
- tubing connected to the wrong valve ports
- a bypassed column or injection loop
- a valve connected to the wrong channel
- disconnected or blocked tubing
-
Perform a manual fixed-loop injection of a known standard when the configuration permits.
Use an appropriate known-good standard and manually load/overfill the installed sample loop according to the applicable injection-valve procedure.
Keep the downstream chromatographic conditions unchanged:
Injection valve → column → suppressor → detector
Compare the result with the failed automated injection.
-
Interpret the manual-injection result.
If the manual injection produces normal peaks:
The downstream chromatographic system is capable of producing a response.
Move suspicion upstream toward:
- autosampler sample delivery
- sample-transfer tubing
- vial/sample position
- sample depletion
- autosampler injection settings
- wash or transfer path
- sampler-to-injection-valve connection
Continue troubleshooting according to the installed autosampler architecture.
If the manual injection still produces no peaks:
Reduce the probability of the autosampler being the root cause.
Continue troubleshooting:
- injection valve
- sample loop
- valve plumbing
- column/flow path
- suppressor
- detector
- Chromeleon acquisition/configuration
-
For an AS-AP, identify the actual injection mode before troubleshooting sample delivery.
Determine whether the AS-AP is configured for the applicable Push or Pull architecture and whether the method uses full-loop, partial-loop, concentrator, or another supported injection mode.
When appropriate, full-loop operation can be used as a troubleshooting simplification.
Consider:
- syringe condition and bubbles
- needle/injection-port alignment
- vial/sample volume
- buffer and transfer tubing
- sample-transfer-line volume where applicable
- valve operation
- sample aspiration/delivery
- wash configuration
The AS-AP supports multiple injection architectures and should not be treated as a generic autosampler. Man-IC-AS-AP-Ops-July2025-DOC06…
-
For an AS-DV, troubleshoot its displacement/full-loop delivery architecture rather than partial-loop metering.
Verify:
- vial fill
- PolyVial cap condition and installation
- sampling-tip condition
- sample-out tubing
- loop volume
- delivered volume
- delay volume
- flush factor
- delivery speed
- vial-memory/reset condition
- optional AS-DV valve path, if installed
The AS-DV uses the vial cap as an active piston to displace sample through the sample-out line. AS-DV_Service_Manual_Rev_2
-
Confirm injection-valve actuation and plumbing if manual injection does not produce peaks.
Command the applicable valve and verify that it reaches the expected Load and Inject positions.
Confirm:
- correct valve assignment
- correct sample-loop connections
- correct analytical inlet/outlet connections
- no crossed ports after maintenance
- no leak at the valve or loop
-
Evaluate the column and analytical flow path.
Do not assume that a bad column is the primary cause simply because peaks are absent.
Compare:
- current versus normal pressure
- retention behavior from the last successful run
- recent column or guard replacement
- recent replumbing
- known-good standard performance
An abrupt loss of all peaks with otherwise normal pressure should keep injection, valve, detector, and software/configuration high in the fault tree.
-
Evaluate suppression when suppressed conductivity detection is used.
Check:
- suppressor on/off state
- applicable suppressor current or operating state
- regenerant or external-water flow where applicable
- suppressor chemistry and configuration
- bubbles or outgassing
- background conductivity
- downstream restriction
Do not replace the suppressor solely because chromatographic peaks are absent.
A gross suppression problem usually provides additional evidence such as abnormal background, noise, poor response, unstable conductivity, or an error condition.
-
Evaluate detector response.
If flow reaches the detector but no injected standard response is observed, investigate the detector branch.
For a conductivity detector, inspect:
- detector channel acquisition
- conductivity background
- detector-cell flow
- bubbles
- cell condition
- temperature where applicable
- downstream outlet flow/restriction
Detector testing should be separated conceptually from injector and pump testing. Thermo's integrated IC qualification procedure independently evaluates baseline noise/drift, detector linearity, injector repeatability, and pump-flow performance. [Procedures]CDS OQ Chromeleon A…
-
Use a simplified Chromeleon method if hardware operation appears normal.
Create or use a known-good diagnostic Instrument Method containing only the commands necessary for:
- pump flow
- detector acquisition
- required suppression/electrolytic devices
- injection
Run a known standard.
If the simplified method produces peaks but the customer method does not, troubleshoot the original Instrument Method, sequence, channel assignment, valve commands, timing, or configuration instead of replacing hardware.
-
Verify the repair or correction using the original method.
After identifying and correcting the fault:
- rerun the original standard
- confirm expected pressure and flow
- confirm expected detector baseline
- verify retention time
- verify peak area and height
- verify repeatability
- confirm no new Audit Trail errors
NOTE: Do not consider the problem resolved only because a simplified diagnostic method works.
Analysis