Troubleshoot a rear seal-wash peristaltic pump that is not operating on a Dionex ICS-5000 Plus DP
Issue
The rear seal-wash peristaltic pump on a Thermo Scientific Dionex ICS-5000+ DP does not appear to operate.
The rear seal-wash pump may not rotate when commanded, or it may rotate without moving seal-wash liquid.
Environment
- Product: Thermo Scientific ICS-5000+ IC System
- Product: Thermo Scientific ICS-5000+ SP (single pump) or ICS-5000+ DP (dual pump) System
- Software: Thermo Scientific Chromeleon CDS
Resolution
NOTE: This article applies specifically to the rear pump seal-wash system. Do not assume that a rear seal-wash failure indicates a failure of the analytical pump heads, pistons, or high-pressure pump seals.
Recommended Troubleshooting Steps
-
Confirm that the rear seal-wash pump is being commanded to operate.
Use the applicable Chromeleon instrument controls or F8 Commands and verify that the rear seal-wash function is enabled and commanded ON.
Do not replace the seal-wash pump until it has been confirmed that the pump is actually being commanded.
-
Observe the rear seal-wash pump while it is commanded ON.
Determine whether the peristaltic pump rotor and rollers rotate.
At the same time, observe the seal-wash tubing for liquid movement.
-
If the pump rotates but no liquid moves, troubleshoot the seal-wash fluid path before replacing the pump.
Verify:
- The seal-wash reservoir contains fresh DI water.
- The inlet tubing is submerged in the reservoir.
- Tubing is correctly installed in the peristaltic pump.
- Tubing is not kinked, collapsed, pinched, disconnected, or blocked.
- The pump tubing is correctly captured by the rollers.
- The outlet tubing and pump-head seal-wash path are not obstructed.
- Connections at the pump-head seal-wash bodies are correctly installed.
-
If the pump does not rotate, inspect the pump electrical connection.
Place the instrument in the appropriate safe service state before accessing internal components.
Inspect the rear seal-wash pump connector and wiring for:
- A loose or partially seated connector
- Bent or damaged connector pins
- Broken or pulled wires
- Cable damage
- Contamination or corrosion
- Intermittent operation when the connector or cable is moved
-
Determine whether the pump is receiving the specified electrical drive when commanded ON.
Use the applicable ICS-5000+/ICS-6000 service documentation for the correct connector, pin assignment, and electrical specification.
Do not assume a generic motor voltage.
-
Interpret the result before replacing parts.
- Correct electrical drive is present, but the pump does not rotate: the rear seal-wash pump assembly is strongly indicated as failed.
- No electrical drive is present at the pump when commanded ON: do not replace the pump yet. Continue troubleshooting the cable, connector, pump-control circuitry, and associated electronics.
- Pump rotates normally but no liquid moves: continue troubleshooting tubing and the seal-wash fluid path.
- Pump operates intermittently when wiring is moved: investigate the electrical connection or harness before replacing the pump.
-
Replace the rear seal-wash pump only after the pump itself has been isolated as the failed component.
Replace the rear seal-wash peristaltic pump assembly with:
P/N 062058 – ASSY, SEAL WASH, PMP, ICS3/5
Follow the applicable ICS-5000+/ICS-6000 service-manual procedure for access, removal, mounting, tubing orientation, and electrical reconnection.
-
Do not replace unrelated pump-head components solely because the seal-wash pump failed.
The seal-wash pump, seal-wash tubing, seal-wash body, analytical pump seals, and analytical pump components are separate service items.
Replace additional components only when inspection or testing provides evidence that they are defective.
-
Verify the repair before returning the instrument to service.
Command the rear seal-wash pump ON and verify:
- The pump starts reliably.
- The rotor turns normally.
- Seal-wash liquid is drawn from the reservoir.
- Liquid moves through the seal-wash circuit.
- There are no leaks.
- Tubing remains correctly positioned in the peristaltic pump.
- The pump does not exhibit abnormal noise.
- Repeated ON/OFF commands produce repeatable operation.
-
Return the analytical pump to normal operation and perform a final system check.
Verify:
- Normal pump pressure
- Stable pressure behavior
- No new leak condition
- No visible leakage around the pump-head/seal-wash area
- Normal operation under the customer's original method
A rear seal-wash pump replacement by itself does not justify changing analytical pump calibration.
Analysis