Troubleshoot an ICS-4000 Sequence That Repeatedly Stops During Analysis
Issue
A Thermo Scientific Dionex ICS-4000 sequence repeatedly stops or aborts during analysis.
The interruption may occur at approximately the same point in the chromatogram, for example after the chloride peak, or may occur intermittently during different injections.
The cause may not be obvious from the chromatogram alone. Possible fault domains include:
- eluent generator or other electrolytic-device condition
- suppressor condition
- pump, flow, or pressure problem
- conductivity detector problem
- Chromeleon instrument method or sequence command
- external control or communication event
Environment
- Product: Thermo Scientific Dionex ICS-4000
- Software: Thermo Scientific Chromeleon CDS
Resolution
NOTE: Record the exact instrument configuration, Chromeleon version, method conditions, installed electrolytic devices, and exact Audit Trail message before applying product-specific corrective actions.
Recommended Troubleshooting Steps
-
Determine what Chromeleon means by the sequence "stopping."
Reproduce the problem with the Chromeleon Audit Trail, Queue, and instrument status visible.
At the time of the interruption, record:
- exact time of the event
- injection number
- queue status
- exact Audit Trail message
- error severity
- last completed instrument command
- system pressure
- conductivity
- pump state
- eluent generator state/current, if applicable
- suppressor state/current, if available
- detector state
Do not replace any components yet.
-
Identify the first abnormal event.
Compare the Audit Trail and instrument traces at the exact time the sequence stops.
Determine whether the first abnormal event is:
- an EGC/electrolytic-device error
- a suppressor error
- a pressure or flow disturbance
- a detector error
- a Chromeleon Abort/Stop command
- no hardware error at all
The first event is generally more diagnostically useful than secondary errors that occur after flow or device operation has already been interrupted.
-
Evaluate the EGC/eluent-generation branch when applicable.
If an EGC error occurs, determine whether the EGC condition occurred before or after a pressure or flow disturbance.
- If the EGC state/current changes first while pressure remains normal, continue troubleshooting the eluent-generation/electrolytic-device path.
- If pressure or flow changes first and the EGC error follows, troubleshoot the hydraulic/flow problem before replacing the EGC.
A downstream EGC error can be a reaction to loss of valid flow rather than the initiating failure.
-
Evaluate the suppressor branch.
Move the suppressor higher in the fault tree when:
- pump pressure remains stable
- flow appears stable
- EGC operation remains stable
- suppressor current or status changes
- conductivity becomes abnormal
- an Audit Trail message identifies the suppressor
Check suppression conditions, regenerant or external-water delivery as applicable, bubbles/outgassing, suppressor state, and downstream detector/outlet conditions.
Do not assume a suppressor leak or suppressor error proves that the suppressor itself is the root cause. Downstream restriction should also be considered where applicable.
-
Evaluate the pump and hydraulic branch.
Examine the pressure trace immediately before and during the interruption.
Determine whether pressure:
- remains stable
- drops
- rises
- becomes erratic
- falls to zero when the sequence stops
If pressure changes before other device errors, prioritize pump/flow/fluidic troubleshooting.
If commanded flow is in question, measure actual collected flow using an appropriate verified procedure.
Do not adjust pump calibration to compensate for an unresolved hydraulic problem.
-
Evaluate the detector branch.
Suspect the detector more strongly when:
- pressure remains stable
- flow remains stable
- EGC and suppressor states remain normal
- the detector itself produces an error or Abort condition
- conductivity becomes physically abnormal before the queue stops
If detector signal changes only after flow or suppression has already failed, continue troubleshooting the upstream failure first.
-
Determine whether the interruption occurs at the same elapsed time or merely near the same chromatographic peak.
If the run repeatedly stops at the same elapsed time, inspect the instrument method for a timed event at that point, including:
- EGC concentration changes
- suppressor commands
- valve commands
- pump commands
- detector commands
- temperature commands
- TTL/relay events
- Stop or Abort-related commands
If the failure consistently occurs "after chloride," compare the chloride retention time with the actual instrument-method timeline. The chloride peak itself may only be acting as a visual time marker for a programmed event.
-
Inspect the raw Chromeleon instrument method.
Review the commands immediately before and after the failure time.
Confirm that the method does not unintentionally:
- turn off the pump
- change flow
- turn off or alter an electrolytic device
- switch a valve
- change detector state
- execute a stop/abort command
- respond to an external TTL input
Also confirm that the sequence is using the intended instrument method for the affected injections.
-
Use a simplified run to separate hardware from sequence control.
When safe and appropriate, run the system under stable direct-control conditions and perform a single test injection or a short sequence of identical standards.
Remove nonessential method events from the diagnostic method.
Monitor:
- pressure
- conductivity
- EGC state/current
- suppressor state
- detector state
- Audit Trail
- queue status
If the system remains stable under direct control but repeatedly stops when the original sequence runs, move Chromeleon method/sequence/control logic higher in the fault tree.
If the same failure occurs outside the sequence, move the relevant hardware/device branch higher.
-
Verify the correction under the original method.
After the initiating cause is identified and corrected, rerun the original method and confirm:
- sequence completes normally
- pressure remains stable
- conductivity is appropriate
- electrolytic devices remain in the expected state
- detector remains operational
- retention and response are acceptable
- no new Audit Trail errors are generated
Analysis