Troubleshoot an early negative dip/system peak, baseline drift, and poor low-level calibration response in ion chromatography
Issue
An ion chromatography chromatogram shows one or more of the following symptoms:
- A negative dip or system peak near the beginning of the chromatogram.
- Baseline drift later in the run.
- Poor response from low-level standards.
- Low calibration points fail acceptance criteria or cause poor calibration performance.
- Early-eluting analytes are difficult to quantify because they are near the negative dip or system disturbance.
The symptoms may result from sample introduction, injection-solvent mismatch, eluent or water contamination, inadequate equilibration, suppression, detector behavior, temperature, or the analytical method. Do not assume the suppressor, detector, or autosampler is defective based on these symptoms alone.
Environment
Thermo Scientific Dionex integrated ion chromatography systems using conductivity detection.
- Product: Thermo Scientific Dionex Inuvion IC Product Family:
- Dionex Inuvion Core
- Dionex Inuvion
- Dionex Inuvion RFIC
- Product: Thermo Scientific Dionex IC Samplers:
- Dionex AS-AP
- Dionex AS-DV
- Manual/fixed-loop injection
Resolution
NOTE: For AS-AP troubleshooting, identify the configured injection mode, including full-loop versus partial-loop operation. For AS-DV, do not use partial-loop logic; instead evaluate loop volume, delivered volume, delay volume, flush factor, delivery speed, vial/cap condition, and the sample-out path.Recommended Troubleshooting Steps
-
Record the current method and system conditions before changing anything.
Record:
- Exact IC platform and configuration.
- Autosampler or manual injection configuration.
- Isocratic or gradient operation.
- Eluent type and concentration.
- Manually prepared versus RFIC-generated eluent.
- Column and guard column.
- Flow rate.
- Expected and actual system pressure.
- Background conductivity.
- Column and detector/cell temperature.
- Injection mode and volume.
- Peak area, peak height, width, and retention time.
- Which analytes are affected.
- Whether the problem appeared suddenly, progressively, or after a recent change.
-
Run the method without making an injection.
Allow the system to operate through the portion of the run where the baseline normally drifts.
- If the baseline drifts without an injection, move suspicion away from the autosampler and injection event. Investigate eluent/water quality, equilibration, suppression, detector behavior, temperature, bubbles, and hydraulic stability.
- If the baseline remains stable until an injection is made, prioritize injection-solvent mismatch, sample introduction, matrix effects, carryover, water-dip behavior, and early system peaks.
-
Determine whether the early negative feature is an injection or void effect.
Compare the current low-level standard with a standard prepared using the diluent specified by the applicable method or column/application guidance.
For carbonate-based methods, water or a significantly different sample diluent can produce an early water dip that interferes with early-eluting analytes. Low-level analytes near this disturbance can appear to have poor response even when the detector is functioning normally.
Overlay the two chromatograms and compare:
- Negative-dip size.
- Early peak area and height.
- Peak shape.
- Retention time.
- Integration behavior.
-
Run repeated injections of one stable standard.
Run approximately 6 to 10 consecutive injections of the same standard and trend:
- Peak area.
- Peak height.
- Peak width.
- Retention time.
- Background conductivity.
- Pressure.
Look for a progressive trend rather than evaluating only one failed standard.
A repeated-standard sequence is a project field troubleshooting practice used to convert an intermittent or sequence-dependent complaint into a time-series diagnostic. It is not a Thermo acceptance specification. Inuvion_Expert_Learned_Knowledg…
-
Separate sample-introduction variability from downstream chromatography.
For an AS-AP, simplify to full-loop operation when practical.
If sample-introduction variability remains suspected, bypass the autosampler where appropriate and use a manual fixed-loop injection.
- Repeatable manual fixed-loop injections move suspicion toward the autosampler/sample-transfer path.
- Continued variability with manual injection keeps the injection valve, column, suppression, detector, eluent, and other downstream branches open.
A manual fixed-loop test removes the autosampler delivery path but does not remove the chromatograph injection valve, column, suppressor, or detector. Integrated_IC_Expert_Architectu…
-
Evaluate peak area, height, and width separately.
Do not diagnose sensitivity loss from peak height alone.
- Lower height with approximately stable area and increased width indicates broadening rather than loss of analyte mass.
- Lower area and lower height with stable retention is more consistent with reduced injected mass or reduced detector response.
- A change affecting all analytes together suggests a global system effect.
- A problem limited to one early analyte suggests water-dip interference, selectivity, coelution, matrix, or integration before a general detector problem.
-
Verify adequate system equilibration.
Monitor:
- Background conductivity.
- Pressure.
- Column temperature.
- Conductivity-cell temperature.
Do not evaluate low-level calibration performance until these values are stable.
The Inuvion operating manual directs the user to equilibrate the system and monitor operating readiness before analysis; its troubleshooting section also identifies insufficient equilibration and temperature changes as contributors to baseline drift. Dionex Inuvion Operating Manual
-
Check eluent preparation and water quality.
For manually prepared eluent:
- Inspect the eluent reservoir.
- Rinse the reservoir and pickup hardware if contamination is suspected.
- Prepare fresh eluent using appropriate Type I DI water and clean labware.
- Inspect the eluent inlet/end-line filter.
- Do not continually top off old eluent without routine reservoir cleaning.
Thermo documentation requires high-purity Type I water for IC work and identifies contaminated or incorrectly prepared eluent as a potential source of baseline problems. Dionex Inuvion Operating Manual
-
Evaluate suppressor performance only after the injection and eluent branches have been considered.
Check:
- Suppressor state.
- Correct suppressor chemistry/mode.
- Suppressor current where applicable.
- Regenerant or external-water flow where applicable.
- Background conductivity.
- Bubbles or outgassing.
- Downstream restriction or excessive backpressure.
Rising background conductivity accompanied by falling analyte response increases suspicion of inadequate suppression.
Stable background with a problem confined to an early low-level analyte near the water dip makes a general suppressor failure less likely.
-
Check detector and temperature behavior.
Trend actual detector/cell and column temperatures rather than relying only on setpoints.
Correlate any baseline movement with:
- Cell temperature.
- Column temperature.
- Ambient changes.
- Pressure.
- Conductivity.
Conductivity is temperature dependent, so temperature instability can appear as baseline drift or poor low-level reproducibility.
-
Review the analytical method before replacing hardware.
Determine whether the calibration problem is:
- Limited to the lowest standard.
- Limited to one early analyte.
- Present for all analytes.
- Associated with a large water/system dip.
- Associated with peak broadening.
- Associated with retention shifts.
If only the lowest standard fails and the affected analyte elutes near the system dip, investigate method/diluent/integration effects before concluding that the detector is nonlinear.
-
Verify the correction using the original analytical method.
After identifying and correcting the cause, confirm:
- Stable baseline.
- Normal background conductivity.
- Stable pressure.
- Stable temperatures.
- Acceptable peak area repeatability.
- Acceptable retention-time repeatability.
- Acceptable low-level calibration response.
- No abnormal system peak or water-dip interference with the analyte of interest.
Analysis