Troubleshoot blue or sticky liquid collecting under the carousel of a Dionex AS-AP Autosampler
Issue
Blue, colored, sticky, or otherwise unidentified liquid collects under or around the sample carousel/tray of a Thermo Scientific Dionex AS-AP Autosampler.
The source of the liquid is not immediately apparent and may appear to originate from beneath the carousel even when the actual leak originates elsewhere in the autosampler.
Environment
- Product: Thermo Scientific Dionex AS-AP Autosampler
Resolution
NOTE: The AS-AP contains several independent liquid paths, including the wash-liquid line, syringe and syringe valve, syringe waste line, buffer line, sampling needle, wash port, injection port, sample-transfer line, drain system, and optional valves. AS-AP systems equipped with sample-temperature control also contain a separate coolant circuit associated with the carousel assembly.
Recommended Troubleshooting Steps
-
Confirm the exact calibration failure before changing hardware.
Review the calibration table and identify:
- Which analyte is failing: nitrate, nitrite, or both
- Which calibration level or levels are abnormal
- Actual coefficient of determination
- Expected versus actual peak area
- Whether the failure is repeatable
Do not diagnose the cause from r² alone.
-
Review every raw calibration chromatogram.
For each level, compare:
- Peak area
- Peak height
- Peak width
- Peak shape
- Retention time
- Baseline before and after each peak
- Peak identification
- Integration start and stop points
Chromeleon quantification depends on correct peak identification and integration. A single incorrectly integrated calibration point can make a physically sound calibration appear nonlinear.
-
Determine whether the method is isocratic or gradient.
For an isocratic method, expect a reasonably stable baseline after adequate equilibration.
For a gradient method, baseline movement may be normal. Compare early and late injections and determine whether the gradient baseline profile is reproducible from run to run.
-
Check pressure stability and rule out trapped air.
Observe pressure under normal method conditions.
Look for:
- Pressure cycling
- Sudden pressure drops
- Pressure substantially below the normal method value
- Pressure instability occurring at the same time as conductivity disturbances
If trapped air is suspected, prime the analytical pump according to the Inuvion operating procedure, allow the system to re-equilibrate, and repeat a mid-level standard.
The Inuvion operating manual identifies inadequate priming and trapped gas as causes of erratic pressure/flow and chromatographic instability. Dionex Inuvion Operating Manual
-
Inspect for leaks before replacing components.
Inspect:
- Pump fittings
- Injection-valve connections
- Column and guard fittings
- Suppressor connections
- Conductivity-cell fittings
- Reaction or post-column tubing, if installed
- Waste and outlet tubing where relevant
Watch pressure and conductivity together. A disturbance in both channels raises the probability of a hydraulic event.
-
Establish whether the baseline is stable without an injection.
Allow the system to equilibrate and observe the conductivity trace without injecting sample.
If the baseline is unstable without injections, investigate:
- Air or incomplete priming
- Leaks
- Eluent contamination or incorrect composition
- Suppression/regeneration
- Bubbles
- Temperature instability
- Conductivity-cell condition
- Insufficient equilibration
Thermo's Inuvion troubleshooting section identifies leaks, trapped gases, eluent condition, temperature, suppressor conditions, and equilibration among causes of baseline noise or drift. Dionex Inuvion Operating Manual
-
Run repeated injections of the same mid-level standard.
Run approximately five to six consecutive injections of one stable nitrate/nitrite standard.
Trend:
- Peak area
- Peak height
- Peak width
- Retention time
- Baseline
- Pressure
Interpret the result:
- Stable area and stable retention time: move suspicion toward calibration standards, concentration assignment, integration, or calibration settings.
- Variable area with stable retention time: sample introduction rises in probability.
- Variable retention time with changing pressure: hydraulic delivery, trapped air, leaks, or flow instability rise in probability.
- Variable retention time with stable pressure: eluent composition, temperature, equilibration, or column chemistry rise in probability.
- Stable repeated standard but nonlinear multi-level calibration: standards, integration, concentration assignment, overload, or analyte chemistry become more likely than hardware failure.
Repeated identical standards are an established Integrated IC troubleshooting method because they convert a sequence complaint into a controlled time-series experiment. Inuvion_Expert_Learned_Knowledg…
-
Inspect reaction-cell or post-column tubing only if the application actually uses it.
The standard Inuvion flow path does not include a component generically identified as a "reaction cell."
If the application includes additional post-column reaction tubing or an external reaction device, inspect for:
- Bubbles
- Loose fittings
- Leaks
- Kinks
- Partial restriction
- Excessive dead volume
- Poorly seated tubing
- Changes in tubing length or internal diameter
If the application permits, bypass the reaction assembly and repeat the standard.
If calibration performance improves when the reaction path is removed, continue troubleshooting that application-specific assembly.
-
Evaluate chromatography before changing Chromeleon calibration settings.
Determine whether retention shifts affect:
- Both nitrate and nitrite
- All analytes in the chromatogram
- Only one analyte
A global retention shift points first toward:
- Actual flow
- Eluent concentration/composition
- Eluent-generation output where applicable
- Temperature
- Equilibration
- Broad column effects
An analyte-specific problem raises:
- Selectivity
- Matrix effects
- Coelution
- Standard stability
- Integration
- Analyte-specific chemistry
-
Check nitrite standard stability.
Nitrite deserves additional attention because it can oxidize to nitrate.
If nitrate calibrates normally while nitrite response degrades or becomes inconsistent, inspect:
- Age of standards
- Standard preparation
- Storage conditions
- Time between preparation and analysis
- Possible nitrite-to-nitrate conversion
Thermo's Inuvion operating guidance notes nitrite oxidation to nitrate as an analytical consideration. Dionex Inuvion Operating Manual
- Verify calibration standards and concentration assignments.
Confirm:
- Correct standard concentration at every level
- Correct units
- Correct vial/sequence assignment
- Correct component assignment
- Correct actual concentration entered in the quantification method where certified standards are used
- Freshly prepared standards where required
Thermo's integrated IC OQ procedure instructs entering actual certified standard concentrations into the quantification files when applicable. [Procedures]CDS OQ Chromeleon A…
- Review Chromeleon processing and calibration settings.
Verify:
- Nitrate and nitrite are correctly identified
- Retention windows are appropriate
- Peak integration is correct
- No inappropriate manual integration is applied to one calibration level
- Correct processing method is assigned
- Correct calibration model is selected
- Correct units and level assignments are used
- Any weighting or forced-zero setting is intentional and method-approved
- No calibration level is accidentally excluded or duplicated
Do not delete a failing calibration point solely to improve r² without determining why the point failed.
- Do not recalibrate the conductivity detector to correct an analytical calibration problem.
Conductivity-cell calibration is a separate service function.
Do not use detector calibration, pump calibration, or other instrument calibration adjustments to compensate for:
- Leaks
- Air
- Flow instability
- Poor standards
- Bad peak integration
- Reaction-tubing problems
- Chromatographic instability
The Integrated IC project specifically requires hydraulic and analytical faults to be corrected before calibration adjustments are considered. Integrated_IC_Expert_Master_Dir…
- Verify the correction under the original method.
After identifying and correcting the cause:
-
Do not replace the syringe, syringe valve, tubing, or injection-port components until the leak source is localized.
-
Stop the sequence and clean all visible liquid from the autosampler. Dry the underside of the carousel, syringe area, wash/injection-port area, tubing, and accessible drain surfaces.
-
Record the characteristics of the liquid:
- color
- whether it is watery or sticky/viscous
- approximate quantity
- whether it appears continuously or only during an autosampler operation
Do not identify the liquid solely from its color.
-
Determine whether the AS-AP has the sample-temperature-control option.
If temperature control is installed:
- turn sample temperature control off;
- dry the affected area;
- allow the autosampler to remain idle and check for additional liquid;
- then operate the temperature-control system and observe whether the leak returns.
If the leak occurs preferentially while the temperature-control system is operating, inspect the carousel coolant circuit before assuming the liquid is wash or sample fluid.
The AS-AP temperature-controlled carousel contains coolant plumbing and coolant connections associated with the carousel assembly. 065437_AS-AP_Service Manual_Rev…
-
Place clean absorbent material at several separated locations, where practical:
- below the syringe/syringe-valve area;
- below the wash/injection-port area;
- beneath or at the lowest accessible point of the carousel area.
Use the first location that becomes wet to help identify the source. Do not diagnose from the final puddle location alone because liquid may travel along tubing or chassis surfaces before collecting.
-
Test the syringe and syringe-valve path.
Dry the syringe and syringe-valve fittings. Run a manual syringe prime while observing:
- syringe body and plunger;
- syringe-to-valve connection;
- wash-reservoir-line connection;
- buffer-line connection;
- syringe waste-line connection.
The AS-AP troubleshooting documentation identifies a cracked syringe, leaking plunger seal, worn or missing syringe-valve gasket, leaking fittings, and damaged liquid lines as potential leak sources. Man-IC-AS-AP-Ops-July2025-DOC06…
-
Test the wash-liquid path independently.
Run a manual wash cycle and observe:
- wash-reservoir tubing;
- syringe valve;
- buffer tubing;
- sampling needle;
- wash port;
- associated waste/drain path.
If liquid first appears during a manual wash but not while the system is idle, move the wash-fluid or waste path higher in the diagnostic priority.
-
Inspect the syringe waste and drain system.
Check for:
- loose or damaged waste tubing;
- kinked or blocked drain tubing;
- poor drain-line routing;
- liquid backing up into the autosampler;
- a drain-line connection leak.
Thermo AS-AP troubleshooting specifically identifies leaking drain-line connections as a liquid-leak source. Man-IC-AS-AP-Ops-July2025-DOC06…
-
Inspect the wash port and needle alignment.
During a manual wash, observe whether liquid escapes around the wash port rather than draining normally.
If liquid appears around the port, inspect needle alignment and the wash-port area before replacing unrelated tubing.
-
Test the injection path separately.
Perform a controlled injection while observing the injection port and sample-transfer path.
In Push-mode configurations, inspect the path from the needle through the injection port and sample-transfer tubing to the IC injection valve.
If liquid appears only during the injection/load portion of the cycle, prioritize:
- injection-port fitting;
- sampling needle;
- needle depth/alignment;
- sample-transfer tubing;
- fittings in the sample-transfer path.
Thermo's AS-AP troubleshooting information identifies leaking fittings, damaged or worn sampling needles, needle-alignment/depth issues, and sample-transfer-path problems as possible causes of leakage at the injection port. Man-IC-AS-AP-Ops-July2025-DOC06…
-
If the AS-AP contains an optional high-pressure valve, inspect the valve only after determining whether liquid first appears at that valve.
Check:
- tubing connections;
- fittings;
- the front/stator area;
- evidence of leakage from behind the stator.
A high-pressure valve leak is a documented AS-AP liquid-leak category. 065437_AS-AP_Service Manual_Rev…
-
Use the operating event that produces the liquid to localize the fault:
| Observation | Prioritize |
|---|---|
| Leak occurs while completely idle | coolant circuit, static tubing/reservoir leak, or residual liquid |
| Leak occurs when temperature control operates | sample-temperature-control coolant circuit |
| Leak starts during syringe prime | syringe, syringe valve, wash-reservoir line, buffer line, syringe waste |
| Leak starts during manual wash | wash line, wash port, needle alignment, waste/drain path |
| Leak starts only during injection | injection port, needle, sample-transfer line, injection plumbing |
| Leak occurs at an optional valve | fitting or high-pressure valve |
| Liquid appears only after multiple cycles | small intermittent leak, overflow, drain restriction, or liquid migrating from another location |
-
After the source is identified and repaired, clean and dry the autosampler and repeat the operation that originally produced the leak.
-
Verify:
- no new liquid accumulation;
- normal syringe prime and wash behavior;
- normal sample loading/injection;
- acceptable injection repeatability;
- no relevant Chromeleon Audit Trail errors.
Analysis