Troubleshoot poor suppressor performance after installation or PM part replacement on an ICS-5000 Plus
Issue
A newly installed or replaced suppressor on a Dionex ICS-5000+ does not perform correctly after installation.
Symptoms may include:
- High background conductivity
- Low analyte response
- Unstable or noisy baseline
- Poor suppression
- Bubbles
- Suppressor leakage
- Unexpected system pressure
- Suppressor current or voltage abnormalities
- Chromatography that has not returned to normal after suppressor replacement
The suppressor should not be assumed defective solely because the problem began after suppressor installation.
Environment
- Thermo Scientific Dionex ICS-5000+ ion chromatography system.
- Suppressed conductivity applications using a Thermo Scientific Dionex suppressor.
Resolution
Note: Use the operating and installation instructions for the exact suppressor model and generation when determining conditioning, regenerant flow, current, pressure, and back-pressure requirements. Do not apply numerical operating limits from another suppressor generation.
Applicable configurations may include:
- Anion or cation suppression
- 2 mm or 4 mm suppressors
- Chemical or electrolytic suppression
- Recycle or external-water regeneration, depending on suppressor type
- Manually prepared or electrolytically generated eluent
-
Identify the exact suppressor configuration.
Record the suppressor model and part number, anion or cation type, 2 mm or 4 mm format, eluent chemistry, analytical flow rate, and suppression/regeneration mode.
Also record:
- Normal and current background conductivity
- Normal and current system pressure
- Suppressor current and voltage, when applicable
- Exact Chromeleon Audit Trail messages
- Whether the problem began immediately after installation or developed later
-
Determine whether the observed problem is actually consistent with suppression failure.
Run a known-good standard and evaluate:
- Background conductivity
- Peak area
- Peak height
- Peak width and shape
- Retention time
- System pressure
High background together with reduced analyte response increases suspicion of suppression or regeneration problems.
Normal background with abnormal chromatography should also prompt investigation of the column, eluent, injection system, flow, temperature, or sample matrix.
-
Verify suppressor plumbing against the installation diagram for the exact suppressor.
Confirm:
- Eluent inlet is connected to the correct suppressor port.
- Eluent outlet is connected toward the detector.
- Regenerant or external-water inlet is connected correctly.
- Regenerant or waste outlet is connected correctly.
- No inlet and outlet lines are reversed.
- No shipping plugs or caps remain installed.
- Tubing is not kinked, crushed, restricted, or incorrectly connected.
- Fittings are properly seated and leak-free.
- The suppressor electrical cable is fully connected, when applicable.
-
Verify that the suppressor was conditioned or hydrated according to the applicable suppressor procedure.
Do not assume that a newly installed suppressor will immediately provide normal background and response before the required startup or conditioning procedure has been completed.
If conditioning was not completed, perform the conditioning procedure specified for that suppressor model and generation.
-
Verify actual regenerant or external-water flow.
Confirm that liquid is physically moving through the regeneration side of the suppressor.
Check for:
- Empty regenerant or water reservoir
- Air in the regenerant line
- Kinked tubing
- Restricted tubing
- Incorrect tubing connections
- Pump or delivery problems
- Restricted waste tubing
- Bubbles entering or leaving the suppressor
Do not rely only on a software command or pump status indication. Confirm actual flow.
-
Verify actual analytical flow.
If analytical flow is questionable, collect the outlet flow for a measured period and compare actual flow with the commanded method flow.
If actual analytical flow is incorrect, troubleshoot the pump and fluidic system before replacing the suppressor.
-
Evaluate system pressure and isolate restrictions.
Compare current pressure with the established normal pressure for the method.
If pressure increased after suppressor replacement, inspect the flow path for restrictions.
Potential locations include:
- Suppressor tubing
- Conductivity cell
- Conductivity-cell outlet tubing
- Waste tubing
- Other downstream fittings or components
Isolate the flow path at appropriate documented connection points and observe the pressure change after each isolation step.
Do not assume that a suppressor leak proves the suppressor caused the failure. Excessive downstream restriction can place abnormal pressure on the suppressor and make the suppressor the visible leak point.
-
Verify suppressor electrical operation in Chromeleon when an electrolytic suppressor is used.
Check:
- Suppressor commanded state
- Actual suppressor current
- Suppressor voltage
- Stability of current during operation
- Audit Trail errors or warnings
If the suppressor is commanded ON but actual current is zero or intermittently drops to zero, investigate the suppressor cable, connector, power/control path, and suppressor electrical continuity before replacing the suppressor.
-
Evaluate the conductivity cell and downstream outlet path.
If analytical flow, regenerant flow, suppressor conditioning, and suppressor electrical operation are normal, evaluate:
- Conductivity-cell contamination
- Bubbles in the cell
- Conductivity-cell restriction
- Cell outlet restriction
- Waste-line restriction
- Detector condition
A downstream detector or outlet problem can produce symptoms that resemble suppressor failure.
-
Run the system without an injection.
Operate the method under normal conditions without injecting a sample.
Monitor:
- Conductivity
- Pressure
- Suppressor current
- Baseline stability
- Regenerant or external-water flow
If the problem is present without injections, the autosampler and sample matrix are less likely causes.
If the problem occurs only after injections, investigate sample introduction, carryover, sample matrix, or contamination.
-
Confirm operation with a known-good standard.
After correcting any installation, flow, electrical, plumbing, or restriction problem, run a fresh known-good standard.
Verify:
- Background conductivity has returned to the expected range.
- Suppressor current and voltage are stable.
- System pressure is normal.
- Peak area, height, width, and retention time are acceptable.
- No suppressor leakage is present.
- Repeatability is acceptable.
-
Replace the suppressor only after the surrounding system has been isolated.
Suppressor replacement is justified when installation, conditioning, analytical flow, regeneration flow, electrical operation, downstream restriction, conductivity-cell condition, and application conditions have been reasonably eliminated as causes.
Analysis