What are the system’s vital signs to monitor to ensure the IC system is operating as intended?
Environment
Ion Chromatography System
Answer
For IC systems with Conductivity Detection, the most common vital signs to monitor are:
- Backpressure
- Background signal
- Baseline noise and drift
- Peak shape
- Retention and peak area repeatability
For IC systems with Electrochemical Detection, the most common vital signs to monitor are:
- Backpressure
- Background signal
- Baseline noise and drift
- Peak shape
- Retention and peak area repeatability
- pH
- Correct waveform selected