Thermo Fisher Scientific
What is an indication that the borate trap needs replacement on an IC system with electrochemical detection running a carbohydrate analysis?
Environment
IC-ED Systems
Carbohydrate analysis 
Answer
Tailing on the mannose peak on the Mix of Six standard.