Thermo Fisher Scientific
What is recommended as the first step when observing inconsistent/non-repeatable results while using an AS-AP sampler?
Environment
AS-AP Autosampler  
Answer
Ensure the AS-AP sampler is fully flushed (i.e. 20 syringe cycles, followed by running the wash fluidics for 5000µL twice) as a starting point.