What is the part number for the ICS-6000 Pump Motor
Issue
How to replace the Pump Motor in the ICS-6000 Pumps.
How to replace the Pump Mechanism in the ICS-6000 Pumps.
How to replace the Pump Drive in the ICS-6000 Pumps.
Note: The "Pump Mechanism" is the official description for this part, but is often referred to as the "Pump Motor" or the "Pump Drive"
Environment
ICS-6000 SP Pump - Analytical and Capillary
ICS-6000 DP Pump - Analytical and Capillary
Part Number
The ICS-6000 Analytical Pump Mechanism is part number is 079649 and the SAP description for this is: "ASSY,PUMP DRIVE,SVC,HPA,ICS5+"
The ICS-6000 Capillary Pump Mechanism is part number is 062060 and the SAP description for this is: "PUMP DRIVE ASSY,SVC,ICS3/5"
Part Image
Resolution
The follow steps are used to replace the Pump Mechanism (Taken from the ICS-6000 DP - Service Manual) :
- Open the prime valve to drop the pump pressure to approximately zero psi.
- Turn off the DP/SP motor in Chromeleon. Wait until the system pressure reaches approximately zero psi.
- Turn off power from the rear main pump power switch. Disconnect the power cord from pump module.
- Unstack the SP or DP pump from the system, remove USB cables and place on a separate bench.
- Disconnect liquid lines.
- Open the front door of the DP/SP chassis.
- Remove the pump heads (refer to the Operator’s Manual, Document No.22181-97002).
- Remove the seal wash mechanism and pistons, and disconnect the solvent lines from the degasser chamber.
- Three (3) screws Bottom and Five on Top (5) connect each of the pump trays in the chassis. Remove the appropriate screws using a Phillips #3 screwdriver refer to “Pump Compartment Screws”.
- Disconnect the cable bundle from the retaining clips. If replacing a capillary pump mechanism, unplug the yellow and black cable (heater cable) from the main board location J20 (that leads to the pump pulse damper).
- Remove the ground wires from chassis mounting tray (upper or lower) depending on which pump mechanism is being replaced.
- From the inside of the pump chassis, loosen the tray lock screw from the retaining bracket, lift, and push the pump tray forward. There is a lock screw on both top and bottom trays.
- If removing the bottom pump tray then loosen the leak sensor by 1/4-turn counterclockwise and raise up slightly to clear the drip tray. Remove front drip tray by removing the two screws that attach the drip tray to the pump tray assembly and let the tray hang down. Do not disconnect the drain line.
- Slide the tray out to expose the pump mechanism. Detach the degasser vacuum tubing. Reference figure for picture of pump mechanism
- Turn the chassis 1/4-turn to expose 4 or 5 Phillips mounting screws at the chassis bottom plate. Remove the screws and set the chassis back upright to remove the pump mechanism.
- Remove the ground wires from the bulkhead panel and pump mechanism. All ground wires must be reinstalled on the new pump mechanism or main board faults can occur.
- Carefully lift the mechanism back and away from the chassis to expose wiring.
- If removing a capillary pump mechanism, then the pump damper mounting bracket must be removed prior to removal of the old pump mechanism.
- Carefully remove the pump pulse damper including the pump damper mounting bracket. Lift the mechanism back and away from the chassis to expose the wiring
- Disconnect the ribbon cable connecting to the CPU board. The ribbon cable is ejected by pushing down on end-clamps
- Turn assembly upside down (rubber feet on top) to expose the power connector.
- A small flat blade screwdriver can be used to push the connector (white in picture) away fromthe board. Note the position of the black and red wires and how the connector is installed before removing the connector
- The power connection (red and black wires) that connect to the Power Supply Board route through the mechanism
- Install the new pump mechanism by performing this procedure in reverse.
- Verify calibration for the new pump mechanism.
- To ensure correct operation of the pump and to avoid operating errors, update or check the following settings in Chromeleon. General instructions for how to enter service modes and set calibration values can be found in “Enable Chromeleon Service Mode” on page 48.
- CamZero: Enter the CamZeroRight and/or CamZeroLeft (=the compensation value for the camshaft zero passage). Enter the value indicated on the pump block label. The factory value should be appropriate for minimum pressure ripple. a. Enter Service Modes b. Open the Commands list by pressing the F8 key on the computer keyboard. Enter Service mode for Control Commands. c. Go to Service Code command and enter in the command window 87794, then click on the Execute button.
- If pressure ripple is found to be higher than expected, troubleshooting the flow components should solve the problem. If this does not suffice, the CamZero value can be adjusted slightly and baseline monitored to optimize the value.
- Set-up Pump to adjust CamZero
- Install a backpressure coil to provide 2000 psi +/-500psi @ 1mL/ min (for analytical)
- Install a backpressure coil to provide 2000 psi +/-500psi @ 0.01mL/min (for capillary)
- Use Chromeleon to acquire pump pressure with the following settings: Pump_1_Pressure.Step = 0.10 [s] Pump_1_Pressure.Average =Off
- Start the pump and allow it to stabilize for 5 - 10 minutes at 1.0 mL/min (for analytical) and 0.01 mL/min (for capillary).
- Optimize CamZero value
- Acquire a pressure trace to measure peak-to-peak ripple. The objective is to achieve a pressure trace as flat as possible and less than ~2bar or 30 psi difference between the minimum and maximum pressure.
- The Cam Zero Calibration Settings for Left and Right pumps are changed by typing values into CamZeroRight or CamZeroLeft and pressing the Execute button. This sets the new (test) value in pump memory, but does not save it in non-volatile EEPROM.
- The default CAM Zero number is 13000 and normally needs to be set between 12000 to 13100.
- Allow at least two minutes when CamZero settings are changed before measuring the resulting pressure trace
- Save Entered Calibration values
- When the pressure trace is within specifications execute the BurnEEProm command. NOTE: This command is required to store the entered value(s) after the Command Function dialog box is closed.
- X Value: The Proportioning Valve Suction Start (X Value) is only relevant to gradient pumps. If the proportioning valve and pump heads are reused from the old pump mechanism, the X Value should still be valid. If necessary, readjust the Suction Start (X Value) (refer to “Readjust the Suction Start (X value)”, Section 2.4.1).
- Pressure Calibration:If the pump heads and transducer are reused from the old pump mechanism, the pump pressure should not need recalibration. If necessary, refer to “Calibrating System Pressure”, Section 2.3.
- Flow Calibration: Pump flow is factory calibrated at 1 mL/min for analytical (or 0.01 mL/min for capillary) at 2000 psi. Once the pump mechanism is assembled and running, the measured flow rates at the test settings are rarely found to be outside the 0.2% range (+/-0.1%). If the measured flow rate is found to be outside of 1.00+/-0.050 mL/min for analytical (or 0.01 mL/min+/- 0.0005 mL/min for capillary, the problem is most likely a small leak or loss of liquid when collecting. Troubleshooting flow components or checking the test set-up should solve the problem. Keep in mind that a 50 µL drop amounts to 0.5% of 1 mL and 1 second amounts to 1.3% of 1 minute. A field test using a manually - acquired volume should use a total volume of 20 - 100 mLs delivered to minimize measurement errors. See “Enter Calibration Values for the New Pump” on for information on entering FlowCalibration.
- Perform the PumpBlockChanged command to update the related service information in Chromeleon.