What is the purpose of an amino trap and when should it be used?
Environment
IC-ED Systems
Carbohydrate analysis
Answer
The amino trap is installed before the guard column and it is used to delay the elution of amino acids when the carbohydrate sample also contains amino acids. If running AAA-Direct for amino acid analysis, a sugar trap can be used to delay the elution of the sugars or carbohydrates.